Search
Search

| Assay Name: | aca-miR-29a-3p |
|---|---|
| miRBase Accession Number: | MI0018840 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGCACCAUUUGAAAUCGGUU |
| Species: | Anolis carolinensis, Callithrix jacchus, Chicken, Cricetulus griseus, Monodelphis domestica, Ornithorhynchus anatinus, Taeniopygia guttata, Tupaia chinensis, Xenopus tropicalis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUUUGAAAUCGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-29a-3p | MIMAT0021912 |
| Callithrix jacchus | cja-miR-29c | MIMAT0039438 |
| Chicken | gga-miR-29a-3p | MIMAT0001096 |
| Cricetulus griseus | cgr-miR-29c-3p | MIMAT0023888 |
| Monodelphis domestica | mdo-miR-29a-3p | MIMAT0004164 |
| Ornithorhynchus anatinus | oan-miR-29a-3p | MIMAT0006807 |
| View More Rows | ||
| Taeniopygia guttata | tgu-miR-29a-3p | MIMAT0014532 |
| Tupaia chinensis | tch-miR-29c-3p | MIMAT0036546 |
| Xenopus tropicalis | xtr-miR-29a | MIMAT0003656 |