Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10703
          Assay Name: dme-miR-9c-5p
          miRBase Accession Number: MI0000413
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Species: Drosophila melanogaster, Aedes aegypti, Anopheles gambiae, Bactrocera dorsalis, Culex quinquefasciatus, Drosophila ananassae, Drosophila erecta, Drosophila grimshawi, Drosophila mojavensis, Drosophila persimilis, Drosophila pseudoobscura, Drosophila sechellia, Drosophila virilis, Drosophila willistoni, Drosophila yakuba
          Product Type: mirVana® miRNA mimic
          Assay ID MC10703

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000014, mir-9

          Mature miRNA Sequence:

          UCUUUGGUAUUCUAGCUGUAGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Drosophila melanogaster dme-miR-9c-5p MIMAT0000392
          Aedes aegypti aae-miR-9c-5p MIMAT0014212
          Anopheles gambiae aga-miR-9c MIMAT0001530
          Bactrocera dorsalis bdo-miR-9c MIMAT0045397
          Culex quinquefasciatus cqu-miR-9-5p MIMAT0014380
          Drosophila ananassae dan-miR-9c MIMAT0008456
          View More Rows
          Drosophila erecta der-miR-9c MIMAT0008554
          Drosophila grimshawi dgr-miR-9c MIMAT0008606
          Drosophila mojavensis dmo-miR-9c MIMAT0008683
          Drosophila persimilis dpe-miR-9c MIMAT0008746
          Drosophila pseudoobscura dps-miR-9c MIMAT0001213
          Drosophila sechellia dse-miR-9c MIMAT0008812
          Drosophila virilis dvi-miR-9c-5p MIMAT0008924
          Drosophila willistoni dwi-miR-9c MIMAT0008992
          Drosophila yakuba dya-miR-9c MIMAT0009112

          Stem-loop Details



          Drosophila melanogaster

          Stem-loop ID dme-mir-9c
          Stem-loop Accession # MI0000413
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUCACGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000392
          miRBase ID: dme-miR-9c-5p
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Aedes aegypti

          Stem-loop ID aae-mir-9c
          Stem-loop Accession # MI0013436
          Stem-loop Sequence
          CUGAAUCUUUGGUAUUCUAGCUGUAGAAUGUUGAGCUUUGUGAUAUGGUAUCUUCUAAAGCUUUAGUACCAGAGGUCCAACUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014212
          miRBase ID: aae-miR-9c-5p
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Anopheles gambiae

          Stem-loop ID aga-mir-9c
          Stem-loop Accession # MI0001635
          Stem-loop Sequence
          UUUCCGGCUGUAUCUUUGGUAUUCUAGCUGUAGAAUGUUGUUUUCAUUGUAAUAUCUCUAAAGCUUUAGUACCAGAGGUCCAACUGGGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001530
          miRBase ID: aga-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Bactrocera dorsalis

          Stem-loop ID bdo-mir-9c
          Stem-loop Accession # MI0037274
          Stem-loop Sequence
          UCUUUGGUAUUCUAGCUGUAGAUUCUUAAAUAUGCUAUCAUCUAAAGCUUUUAUACCAAAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045397
          miRBase ID: bdo-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Culex quinquefasciatus

          Stem-loop ID cqu-mir-9
          Stem-loop Accession # MI0013581
          Stem-loop Sequence
          AUGCGUAUAAUUUCCGGCUGAAUCUUUGGUAUUCUAGCUGUAGAAUGUUGAGCUUGUUACGGUAUCUUCUAAAGCUUUAGUACCAGAGGUCCAACUGGGAAUUGCUUGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014380
          miRBase ID: cqu-miR-9-5p
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila ananassae

          Stem-loop ID dan-mir-9c
          Stem-loop Accession # MI0008993
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUCACGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008456
          miRBase ID: dan-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila erecta

          Stem-loop ID der-mir-9c
          Stem-loop Accession # MI0009097
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUCGCGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008554
          miRBase ID: der-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila grimshawi

          Stem-loop ID dgr-mir-9c
          Stem-loop Accession # MI0009152
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUCUAUAUCUUGUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008606
          miRBase ID: dgr-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila mojavensis

          Stem-loop ID dmo-mir-9c
          Stem-loop Accession # MI0009236
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUUCACAUAUUGUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008683
          miRBase ID: dmo-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila persimilis

          Stem-loop ID dpe-mir-9c
          Stem-loop Accession # MI0009303
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUUUUGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008746
          miRBase ID: dpe-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila pseudoobscura

          Stem-loop ID dps-mir-9c
          Stem-loop Accession # MI0001306
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUUUUGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001213
          miRBase ID: dps-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila sechellia

          Stem-loop ID dse-mir-9c
          Stem-loop Accession # MI0009374
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUCACGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008812
          miRBase ID: dse-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila virilis

          Stem-loop ID dvi-mir-9c
          Stem-loop Accession # MI0009491
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUUUACAGUUUGUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008924
          miRBase ID: dvi-miR-9c-5p
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila willistoni

          Stem-loop ID dwi-mir-9c-1
          Stem-loop Accession # MI0009559
          Stem-loop Sequence
          UUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUUUUGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008992
          miRBase ID: dwi-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Stem-loop ID dwi-mir-9c-2
          Stem-loop Accession # MI0009591
          Stem-loop Sequence
          UUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUUUUGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008992
          miRBase ID: dwi-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA

          Drosophila yakuba

          Stem-loop ID dya-mir-9c
          Stem-loop Accession # MI0009689
          Stem-loop Sequence
          AUUUUUGCUGUUUCUUUGGUAUUCUAGCUGUAGAUUGUUUCGCGCACAUUGUAUAUCAUCUAAAGCUUUUAUACCAAAGCUCCAGCUUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009112
          miRBase ID: dya-miR-9c
          Mature miRNA Sequence: UCUUUGGUAUUCUAGCUGUAGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 000272
          mirVana® miRNA inhibitor : MH10703


          miRBase Alias:

          Drosophila melanogaster: dme-miR-9c (v16)
          Culex quinquefasciatus: cqu-miR-9 (v18)
          Drosophila virilis: dvi-miR-9c (v20)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline