Search
Search

| Assay Name: | dme-miR-9c-5p |
|---|---|
| miRBase Accession Number: | MI0000413 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUUUGGUAUUCUAGCUGUAGA |
| Species: | Drosophila melanogaster, Aedes aegypti, Anopheles gambiae, Bactrocera dorsalis, Culex quinquefasciatus, Drosophila ananassae, Drosophila erecta, Drosophila grimshawi, Drosophila mojavensis, Drosophila persimilis, Drosophila pseudoobscura, Drosophila sechellia, Drosophila virilis, Drosophila willistoni, Drosophila yakuba |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000014, mir-9 |
Mature miRNA Sequence: | UCUUUGGUAUUCUAGCUGUAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-9c-5p | MIMAT0000392 |
| Aedes aegypti | aae-miR-9c-5p | MIMAT0014212 |
| Anopheles gambiae | aga-miR-9c | MIMAT0001530 |
| Bactrocera dorsalis | bdo-miR-9c | MIMAT0045397 |
| Culex quinquefasciatus | cqu-miR-9-5p | MIMAT0014380 |
| Drosophila ananassae | dan-miR-9c | MIMAT0008456 |
| View More Rows | ||
| Drosophila erecta | der-miR-9c | MIMAT0008554 |
| Drosophila grimshawi | dgr-miR-9c | MIMAT0008606 |
| Drosophila mojavensis | dmo-miR-9c | MIMAT0008683 |
| Drosophila persimilis | dpe-miR-9c | MIMAT0008746 |
| Drosophila pseudoobscura | dps-miR-9c | MIMAT0001213 |
| Drosophila sechellia | dse-miR-9c | MIMAT0008812 |
| Drosophila virilis | dvi-miR-9c-5p | MIMAT0008924 |
| Drosophila willistoni | dwi-miR-9c | MIMAT0008992 |
| Drosophila yakuba | dya-miR-9c | MIMAT0009112 |