Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10798
          Assay Name: dme-miR-281-2-5p
          miRBase Accession Number: MI0000370
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Species: Drosophila melanogaster, Bombyx mori, Culex quinquefasciatus, Drosophila ananassae, Drosophila erecta, Drosophila grimshawi, Drosophila mojavensis, Drosophila persimilis, Drosophila sechellia, Drosophila virilis, Drosophila willistoni, Drosophila yakuba, Locusta migratoria, Spodoptera frugiperda, Tribolium castaneum, Triops cancriformis
          Product Type: mirVana® miRNA mimic
          Assay ID MC10798

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000087, mir-46

          Mature miRNA Sequence:

          AAGAGAGCUAUCCGUCGACAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Drosophila melanogaster dme-miR-281-2-5p MIMAT0000349
          Bombyx mori bmo-miR-281-5p MIMAT0007904
          Culex quinquefasciatus cqu-miR-281-5p MIMAT0014358
          Drosophila ananassae dan-miR-281-2-5p MIMAT0009182
          Drosophila erecta der-miR-281-2-5p MIMAT0009183
          Drosophila grimshawi dgr-miR-281-2-5p MIMAT0009184
          View More Rows
          Drosophila mojavensis dmo-miR-281-2-5p MIMAT0009185
          Drosophila persimilis dpe-miR-281-2-5p MIMAT0009186
          Drosophila sechellia dse-miR-281-2-5p MIMAT0009187
          Drosophila virilis dvi-miR-281-2-5p MIMAT0009188
          Drosophila willistoni dwi-miR-281-2-5p MIMAT0009189
          Drosophila yakuba dya-miR-281-2-5p MIMAT0009190
          Locusta migratoria lmi-miR-281-5p MIMAT0010140
          Spodoptera frugiperda sfr-miR-281-5p MIMAT0041898
          Tribolium castaneum tca-miR-281-5p MIMAT0008384
          Triops cancriformis tcf-miR-281-5p MIMAT0041397

          Stem-loop Details



          Drosophila melanogaster

          Stem-loop ID dme-mir-281-2
          Stem-loop Accession # MI0000370
          Stem-loop Sequence
          CGAAUUGUGAAAUGAAGAGAGCUAUCCGUCGACAGUCAAGUUAAGACCGAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAACAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000349
          miRBase ID: dme-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Bombyx mori

          Stem-loop ID bmo-mir-281
          Stem-loop Accession # MI0008348
          Stem-loop Sequence
          UCUGUUAAUGAAGAGAGCUAUCCGUCGACAGUAUUGCUGAUAAACACUGUCAUGGAGUUGCUCUCUUUAUGAACGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007904
          miRBase ID: bmo-miR-281-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Culex quinquefasciatus

          Stem-loop ID cqu-mir-281-1
          Stem-loop Accession # MI0013564
          Stem-loop Sequence
          UUCUGCAGUUUCGAAUGUGAAAAUAAAGAGAGCUAUCCGUCGACAGUAGGGAUAUGAUUCACUGUCAUGGAAUUGCUCUCUUUAUGUACAAUUCGAUAUUCAACAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014358
          miRBase ID: cqu-miR-281-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Stem-loop ID cqu-mir-281-2
          Stem-loop Accession # MI0023547
          Stem-loop Sequence
          UUCUGCAGUUUCGAAUGUGAAAAUAAAGAGAGCUAUCCGUCGACAGUAGGGAUAUGAUUCACUGUCAUGGAAUUGCUCUCUUUAUGUACAAUUCGAUAUUCAACAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014358
          miRBase ID: cqu-miR-281-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila ananassae

          Stem-loop ID dan-mir-281-2
          Stem-loop Accession # MI0008965
          Stem-loop Sequence
          CGAAUAGUGAAAUGAAGAGAGCUAUCCGUCGACAGUCAAGUUAAAACACGGAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAACAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009182
          miRBase ID: dan-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila erecta

          Stem-loop ID der-mir-281-2
          Stem-loop Accession # MI0009040
          Stem-loop Sequence
          CGAAUUGUGAAAUGAAGAGAGCUAUCCGUCGACAGUCAAGUUAAUACCGAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAACAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009183
          miRBase ID: der-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila grimshawi

          Stem-loop ID dgr-mir-281-2
          Stem-loop Accession # MI0009177
          Stem-loop Sequence
          CGAAUAGCGAAAUGAAGAGAGCUAUCCGUCGACAGUCAAGUUAUAUACCGAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAAUAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009184
          miRBase ID: dgr-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila mojavensis

          Stem-loop ID dmo-mir-281-2
          Stem-loop Accession # MI0009200
          Stem-loop Sequence
          CGAAUAGCGAAAUGAAGAGAGCUAUCCGUCGACAGUCAAGUUAUAUACAUAUUUUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAAUAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009185
          miRBase ID: dmo-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila persimilis

          Stem-loop ID dpe-mir-281-2
          Stem-loop Accession # MI0009312
          Stem-loop Sequence
          CGAAUAGUGAAAUAAAGAGAGCUAUCCGUCGACAGUCAAGUUAAUCAGAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAAUAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009186
          miRBase ID: dpe-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila sechellia

          Stem-loop ID dse-mir-281-2
          Stem-loop Accession # MI0009344
          Stem-loop Sequence
          CGAAUUGUGAAAUGAAGAGAGCUAUCCGUCGACAGUCAAGUUAAGACCGAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAACAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009187
          miRBase ID: dse-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila virilis

          Stem-loop ID dvi-mir-281-2
          Stem-loop Accession # MI0009493
          Stem-loop Sequence
          CGAAUAGCGAAAUGAAGAGAGCUAUCCGUCGACAGUCAAGCUAUACAGAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAAUAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009188
          miRBase ID: dvi-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila willistoni

          Stem-loop ID dwi-mir-281-2
          Stem-loop Accession # MI0009561
          Stem-loop Sequence
          CGAAUAGCGAAAUGAAGAGAGCUAUCCGUCGACAGUCCAGUUUAAUACAUCCGAAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAAUAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009189
          miRBase ID: dwi-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Drosophila yakuba

          Stem-loop ID dya-mir-281-2
          Stem-loop Accession # MI0009634
          Stem-loop Sequence
          CGAAUAGUGAAAUGAAGAGAGCUAUCCGUCGACAGUCAAGUUAAUACCAAUUGUAAUACUGUCAUGGAAUUGCUCUCUUUGUAUAAUAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009190
          miRBase ID: dya-miR-281-2-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Locusta migratoria

          Stem-loop ID lmi-mir-281
          Stem-loop Accession # MI0010642
          Stem-loop Sequence
          AAGAGAGCUAUCCGUCGACAGUACAGUGAGUAAACUGUCAUGGAGUUGCUCUCUUUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010140
          miRBase ID: lmi-miR-281-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Spodoptera frugiperda

          Stem-loop ID sfr-mir-281
          Stem-loop Accession # MI0033610
          Stem-loop Sequence
          AUGAAGAGAGCUAUCCGUCGACAGUAUUGCCAGUAAACACUGUCAUGGAGUUGCUCUCUUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041898
          miRBase ID: sfr-miR-281-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Tribolium castaneum

          Stem-loop ID tca-mir-281
          Stem-loop Accession # MI0008925
          Stem-loop Sequence
          UCGAAAAUGAAGAGAGCUAUCCGUCGACAGUAUAUAAUUUACACUGUCAUGGAGUUGCUCUCUUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008384
          miRBase ID: tca-miR-281-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA

          Triops cancriformis

          Stem-loop ID tcf-mir-281
          Stem-loop Accession # MI0033232
          Stem-loop Sequence
          AAGAGAGCUAUCCGUCGACAGUCAAGAAUUCACGAUACUGUCAUGGAGCUGCUCUCUUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041397
          miRBase ID: tcf-miR-281-5p
          Mature miRNA Sequence: AAGAGAGCUAUCCGUCGACAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 001432
          mirVana® miRNA inhibitor : MH10798


          miRBase Alias:

          Drosophila melanogaster: dme-miR-281-2* (v16)
          Bombyx mori: bmo-miR-281* (v18)
          Culex quinquefasciatus: cqu-miR-281 (v18)
          Drosophila ananassae: dan-miR-281-2* (v18)
          Drosophila erecta: der-miR-281-2* (v18)
          Drosophila grimshawi: dgr-miR-281-2* (v18)
          Drosophila mojavensis: dmo-miR-281-2* (v18)
          Drosophila persimilis: dpe-miR-281-2* (v18)
          Drosophila sechellia: dse-miR-281-2* (v18)
          Drosophila virilis: dvi-miR-281-2* (v18)
          Drosophila willistoni: dwi-miR-281-2* (v18)
          Drosophila yakuba: dya-miR-281-2* (v18)
          Locusta migratoria: lmi-miR-281* (v18)
          Trichosporon caseorum: tca-miR-281* (v16)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline