Search
Search

| Assay Name: | dme-miR-281-2-5p |
|---|---|
| miRBase Accession Number: | MI0000370 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGAGAGCUAUCCGUCGACAGU |
| Species: | Drosophila melanogaster, Bombyx mori, Culex quinquefasciatus, Drosophila ananassae, Drosophila erecta, Drosophila grimshawi, Drosophila mojavensis, Drosophila persimilis, Drosophila sechellia, Drosophila virilis, Drosophila willistoni, Drosophila yakuba, Locusta migratoria, Spodoptera frugiperda, Tribolium castaneum, Triops cancriformis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000087, mir-46 |
Mature miRNA Sequence: | AAGAGAGCUAUCCGUCGACAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-281-2-5p | MIMAT0000349 |
| Bombyx mori | bmo-miR-281-5p | MIMAT0007904 |
| Culex quinquefasciatus | cqu-miR-281-5p | MIMAT0014358 |
| Drosophila ananassae | dan-miR-281-2-5p | MIMAT0009182 |
| Drosophila erecta | der-miR-281-2-5p | MIMAT0009183 |
| Drosophila grimshawi | dgr-miR-281-2-5p | MIMAT0009184 |
| View More Rows | ||
| Drosophila mojavensis | dmo-miR-281-2-5p | MIMAT0009185 |
| Drosophila persimilis | dpe-miR-281-2-5p | MIMAT0009186 |
| Drosophila sechellia | dse-miR-281-2-5p | MIMAT0009187 |
| Drosophila virilis | dvi-miR-281-2-5p | MIMAT0009188 |
| Drosophila willistoni | dwi-miR-281-2-5p | MIMAT0009189 |
| Drosophila yakuba | dya-miR-281-2-5p | MIMAT0009190 |
| Locusta migratoria | lmi-miR-281-5p | MIMAT0010140 |
| Spodoptera frugiperda | sfr-miR-281-5p | MIMAT0041898 |
| Tribolium castaneum | tca-miR-281-5p | MIMAT0008384 |
| Triops cancriformis | tcf-miR-281-5p | MIMAT0041397 |