Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10804
          Assay Name: dme-miR-315-5p
          miRBase Accession Number: MI0000427
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Species: Drosophila melanogaster, Aedes aegypti, Anopheles gambiae, Apis mellifera, Culex quinquefasciatus, Daphnia pulex, Drosophila ananassae, Drosophila erecta, Drosophila grimshawi, Drosophila mojavensis, Drosophila persimilis, Drosophila pseudoobscura, Drosophila sechellia, Drosophila simulans, Drosophila willistoni, Drosophila yakuba, Nasonia longicornis, Nasonia vitripennis, Tribolium castaneum
          Product Type: mirVana® miRNA mimic
          Assay ID MC10804

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000141, mir-315

          Mature miRNA Sequence:

          UUUUGAUUGUUGCUCAGAAAGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Drosophila melanogaster dme-miR-315-5p MIMAT0000407
          Aedes aegypti aae-miR-315-5p MIMAT0014262
          Anopheles gambiae aga-miR-315 MIMAT0001521
          Apis mellifera ame-miR-315-5p MIMAT0001487
          Culex quinquefasciatus cqu-miR-315 MIMAT0014365
          Daphnia pulex dpu-miR-315 MIMAT0012660
          View More Rows
          Drosophila ananassae dan-miR-315 MIMAT0008459
          Drosophila erecta der-miR-315 MIMAT0008539
          Drosophila grimshawi dgr-miR-315 MIMAT0008604
          Drosophila mojavensis dmo-miR-315 MIMAT0008687
          Drosophila persimilis dpe-miR-315 MIMAT0008730
          Drosophila pseudoobscura dps-miR-315 MIMAT0001261
          Drosophila sechellia dse-miR-315 MIMAT0008815
          Drosophila simulans dsi-miR-315 MIMAT0008870
          Drosophila willistoni dwi-miR-315 MIMAT0009036
          Drosophila yakuba dya-miR-315 MIMAT0009084
          Nasonia longicornis nlo-miR-315 MIMAT0018433
          Nasonia vitripennis nvi-miR-315 MIMAT0015689
          Tribolium castaneum tca-miR-315 MIMAT0010096

          Stem-loop Details



          Drosophila melanogaster

          Stem-loop ID dme-mir-315
          Stem-loop Accession # MI0000427
          Stem-loop Sequence
          CACUUAUAUAAUUUUGAUUGUUGCUCAGAAAGCCCUCAUUGUUUACCAGUUGGCUUUCGAGCAAUAAUUGAAACCAGAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000407
          miRBase ID: dme-miR-315-5p
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Aedes aegypti

          Stem-loop ID aae-mir-315
          Stem-loop Accession # MI0013467
          Stem-loop Sequence
          AGUGUUUCUAUAAAAUUUUGAUUGUUGCUCAGAAAGCCGUGCAUUUGAAACGGGUCGCUUUCGAGCAGUAAUCAAAGUCAAAUAAAACACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014262
          miRBase ID: aae-miR-315-5p
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Anopheles gambiae

          Stem-loop ID aga-mir-315
          Stem-loop Accession # MI0001626
          Stem-loop Sequence
          UAUAAAAUUUUGAUUGUUGCUCAGAAAGCCGUGUCGAUUAAGCAAUUCGCUUUCGGGCAGUAAUCAAAGUCAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001521
          miRBase ID: aga-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Apis mellifera

          Stem-loop ID ame-mir-315
          Stem-loop Accession # MI0001592
          Stem-loop Sequence
          GCUCUUUAUGCUUUUGAUUGUUGCUCAGAAAGCCUUGAUUAUGAUAUUGGCUUUCGGGCAAUAAUCAUAAUCACGAAAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001487
          miRBase ID: ame-miR-315-5p
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Culex quinquefasciatus

          Stem-loop ID cqu-mir-315
          Stem-loop Accession # MI0013569
          Stem-loop Sequence
          GAGUGUUUCUAUAAAAUUUUGAUUGUUGCUCAGAAAGCCGUGCAUUCGAAACAUUCCGCUUUCGAGCAGUAAUCAGAGUCAAAUAAAACACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014365
          miRBase ID: cqu-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Daphnia pulex

          Stem-loop ID dpu-mir-315
          Stem-loop Accession # MI0012241
          Stem-loop Sequence
          UAUAUAAUAGCUCCCAAUGUUUUUGAUUGUUGCUCAGAAAGCCGUGCCAUUUCACAUUUUUGGCUGACGUCAAAAAAUCACAACUGGCUUUCGAGAAAUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012660
          miRBase ID: dpu-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila ananassae

          Stem-loop ID dan-mir-315
          Stem-loop Accession # MI0008996
          Stem-loop Sequence
          CACUUAUAUAAUUUUGAUUGUUGCUCAGAAAGCCCUUAUAAAUUAUUAGCUGGCUUUCGAGCAAUAAUUGAAACCAAAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008459
          miRBase ID: dan-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila erecta

          Stem-loop ID der-mir-315
          Stem-loop Accession # MI0009080
          Stem-loop Sequence
          CACUUAUAUAAUUUUGAUUGUUGCUCAGAAAGCCCUAUUAUCUAACAGCUGGCUUUCGAGCAAUAAUUGAAACCAGAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008539
          miRBase ID: der-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila grimshawi

          Stem-loop ID dgr-mir-315
          Stem-loop Accession # MI0009150
          Stem-loop Sequence
          CGCUCAUAUAAUUUUGAUUGUUGCUCAGAAAGCCUUUCACUUGACACUUGGCUUUCGAGUAAUAAUCGAAACCAAAUAAGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008604
          miRBase ID: dgr-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila mojavensis

          Stem-loop ID dmo-mir-315
          Stem-loop Accession # MI0009240
          Stem-loop Sequence
          CCCUUAUAUAAUUUUGAUUGUUGCUCAGAAAGCCCGUAAUUUAACUUAACAUGGCUUUCGAGUAAUAAUCGAAACCAAAUAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008687
          miRBase ID: dmo-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila persimilis

          Stem-loop ID dpe-mir-315
          Stem-loop Accession # MI0009283
          Stem-loop Sequence
          CACUUAUCUAUUUUUGAUUGUUGCUCAGAAAGCCCUUAUAAUUAACCAGUUGGCUUUCGAGCAAUUAUCAAAGCCAAAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008730
          miRBase ID: dpe-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila pseudoobscura

          Stem-loop ID dps-mir-315
          Stem-loop Accession # MI0001356
          Stem-loop Sequence
          CACUUAUCUAUUUUUGAUUGUUGCUCAGAAAGCCCUUAUAAUUAACCAGUUGGCUUUCGAGCAAUUAUCAAAGCCAAAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001261
          miRBase ID: dps-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila sechellia

          Stem-loop ID dse-mir-315
          Stem-loop Accession # MI0009377
          Stem-loop Sequence
          CACUUAUAUAAUUUUGAUUGUUGCUCAGAAAGCCCUUAUUGUACCAGUUGGCUUUCGAGCAAUAAUUGAAACCAGAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008815
          miRBase ID: dse-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila simulans

          Stem-loop ID dsi-mir-315
          Stem-loop Accession # MI0009435
          Stem-loop Sequence
          CACUUAUAUAAUUUUGAUUGUUGCUCAGAAAGCCCUUAUUGUUUACCAGUUGGCUUUCGAGCAAUAAUUGAAACCAGAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008870
          miRBase ID: dsi-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila willistoni

          Stem-loop ID dwi-mir-315
          Stem-loop Accession # MI0009604
          Stem-loop Sequence
          CACUUAUAUAAUUUUGAUUGUUGCUCAGAAAGCCCUCAUUACUUUAUCAGUUGGCUUUCGAGCAAUAAUUGAAACCAAAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009036
          miRBase ID: dwi-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Drosophila yakuba

          Stem-loop ID dya-mir-315
          Stem-loop Accession # MI0009685
          Stem-loop Sequence
          CACUUAUAUAAUUUUGAUUGUUGCUCAGAAAGCCCUUACUAUUUAACAGCUGGCUUUCGAGCAAUAAUUGAAACCAGAUAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009084
          miRBase ID: dya-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Nasonia longicornis

          Stem-loop ID nlo-mir-315
          Stem-loop Accession # MI0016674
          Stem-loop Sequence
          GCUCUCUAUGCUUUUGAUUGUUGCUCAGAAAGCCUCGAUAUCGAUACUGGCUUUCGGGCAAUAAUCAUAAUCACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018433
          miRBase ID: nlo-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Nasonia vitripennis

          Stem-loop ID nvi-mir-315
          Stem-loop Accession # MI0014750
          Stem-loop Sequence
          GCUCUCUAUGCUUUUGAUUGUUGCUCAGAAAGCCUCGAUAUCGAUACUGGCUUUCGGGCAAUAAUCAUAAUCACGGGAGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015689
          miRBase ID: nvi-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA

          Tribolium castaneum

          Stem-loop ID tca-mir-315
          Stem-loop Accession # MI0010590
          Stem-loop Sequence
          GUUCCUUUUGCUUUUGAUUGUUGCUCAGAAAGCCGGGUGUUAGAAAGGCUUUCGGGCAAUAAUCAUUUCCAAAGAGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010096
          miRBase ID: tca-miR-315
          Mature miRNA Sequence: UUUUGAUUGUUGCUCAGAAAGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 000325
          mirVana® miRNA inhibitor : MH10804


          miRBase Alias:

          Drosophila melanogaster: dme-miR-315 (v16)
          Achromobacter aegrifaciens: aae-miR-315 (v18)
          Anaerococcus mediterraneensis: ame-miR-315 (v21)
          Drosophila virilis: dvi-miR-315a (v20)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline