Search
Search

| Assay Name: | dme-miR-315-5p |
|---|---|
| miRBase Accession Number: | MI0000427 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAGC |
| Species: | Drosophila melanogaster, Aedes aegypti, Anopheles gambiae, Apis mellifera, Culex quinquefasciatus, Daphnia pulex, Drosophila ananassae, Drosophila erecta, Drosophila grimshawi, Drosophila mojavensis, Drosophila persimilis, Drosophila pseudoobscura, Drosophila sechellia, Drosophila simulans, Drosophila willistoni, Drosophila yakuba, Nasonia longicornis, Nasonia vitripennis, Tribolium castaneum |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-315-5p | MIMAT0000407 |
| Aedes aegypti | aae-miR-315-5p | MIMAT0014262 |
| Anopheles gambiae | aga-miR-315 | MIMAT0001521 |
| Apis mellifera | ame-miR-315-5p | MIMAT0001487 |
| Culex quinquefasciatus | cqu-miR-315 | MIMAT0014365 |
| Daphnia pulex | dpu-miR-315 | MIMAT0012660 |
| View More Rows | ||
| Drosophila ananassae | dan-miR-315 | MIMAT0008459 |
| Drosophila erecta | der-miR-315 | MIMAT0008539 |
| Drosophila grimshawi | dgr-miR-315 | MIMAT0008604 |
| Drosophila mojavensis | dmo-miR-315 | MIMAT0008687 |
| Drosophila persimilis | dpe-miR-315 | MIMAT0008730 |
| Drosophila pseudoobscura | dps-miR-315 | MIMAT0001261 |
| Drosophila sechellia | dse-miR-315 | MIMAT0008815 |
| Drosophila simulans | dsi-miR-315 | MIMAT0008870 |
| Drosophila willistoni | dwi-miR-315 | MIMAT0009036 |
| Drosophila yakuba | dya-miR-315 | MIMAT0009084 |
| Nasonia longicornis | nlo-miR-315 | MIMAT0018433 |
| Nasonia vitripennis | nvi-miR-315 | MIMAT0015689 |
| Tribolium castaneum | tca-miR-315 | MIMAT0010096 |