Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10836
          Assay Name: ath-miR397a
          miRBase Accession Number: MI0001015
          miRBase Version: v22.1
          Chromosome Location: Chr.4: 2625950 - 2626056 [+] on Build TAIR10.1
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Species: Arabidopsis, Arabidopsis lyrata, Brachypodium distachyon, Camelina sativa, Citrus sinensis, Cucumis melo, Fragaria vesca, Glycine max, Linum usitatissimum, Manihot esculenta, Medicago truncatula, Populus trichocarpa, Prunus persica, Rice, Ricinus communis, Salvia sclarea, Sorghum bicolor, Theobroma cacao, Vitis vinifera, Vriesea carinata
          Product Type: mirVana® miRNA mimic
          Assay ID MC10836

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000120, MIR397

          Mature miRNA Sequence:

          UCAUUGAGUGCAGCGUUGAUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Arabidopsis ath-miR397a MIMAT0000946
          Arabidopsis lyrata aly-miR397a-5p MIMAT0017561
          Arabidopsis lyrata aly-miR397b-5p MIMAT0017563
          Brachypodium distachyon bdi-miR397a MIMAT0012180
          Camelina sativa cas-miR397 MIMAT0045348
          Citrus sinensis csi-miR397-5p MIMAT0018465
          View More Rows
          Cucumis melo cme-miR397 MIMAT0026148
          Fragaria vesca fve-miR397 MIMAT0044552
          Glycine max gma-miR397a MIMAT0021627
          Glycine max gma-miR397b-5p MIMAT0021628
          Linum usitatissimum lus-miR397b MIMAT0027181
          Manihot esculenta mes-miR397b MIMAT0045970
          Medicago truncatula mtr-miR397-5p MIMAT0029984
          Populus trichocarpa ptc-miR397a MIMAT0002038
          Prunus persica ppe-miR397 MIMAT0031496
          Rice osa-miR397a MIMAT0000980
          Ricinus communis rco-miR397 MIMAT0014193
          Salvia sclarea ssl-miR397 MIMAT0022518
          Sorghum bicolor sbi-miR397-5p MIMAT0011359
          Theobroma cacao tcc-miR397 MIMAT0020432
          Vitis vinifera vvi-miR397a MIMAT0006561
          Vriesea carinata vca-miR397-5p MIMAT0041015

          Stem-loop Details



          Arabidopsis

          Stem-loop ID ath-MIR397a
          Stem-loop Accession # MI0001015
          Stem-loop Sequence
          UGAAUGAACAUCAUUGAGUGCAGCGUUGAUGUAAUUUCGUUUUGUUUUUCAUUGUUGAAUGGAUUAAAAGAAUUUAUACCAGCGUUGCGCUCAAUUAUGUUUUUCUA
          Chromosome Location Chr. 4 - 2625950 - 2626056 [+] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0000946
          miRBase ID: ath-miR397a
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: Chr. 4 - 2625950 - 2626056 [+] on Build TAIR10.1

          Arabidopsis lyrata

          Stem-loop ID aly-MIR397a
          Stem-loop Accession # MI0014583
          Stem-loop Sequence
          UCCUGGAUUUGAAUGAACAUCAUUGAGUGCAGCGUUGAUGUAAUUUCCUUUUUUUUUCAUUGUUGAAUGGAAUUAAAAGAAUUUACACCGGCGUUGCGCUCAAUUAUGUUUUUCUUAUUUUCAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017561
          miRBase ID: aly-miR397a-5p
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Stem-loop ID aly-MIR397b
          Stem-loop Accession # MI0014584
          Stem-loop Sequence
          CCUGGGUUUGAAUAAACAUCAUUGAGUGCAGCGUUGAUGUAAGUUUACUUAUUUUAUUCCAUUGUUGAAUGAAUUAAAAUGAUACAGAAUACAUCAGCGUUGCAUUCAAUUAUGUUUUUCUAAUUUUCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017563
          miRBase ID: aly-miR397b-5p
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Brachypodium distachyon

          Stem-loop ID bdi-MIR397a
          Stem-loop Accession # MI0011565
          Stem-loop Sequence
          GAAGAGGCGCAAAGGCAUCAUUGAGUGCAGCGUUGAUGAACAGGGGCCAGGCGACCGGCGGCCGGUCCGGUUCGGUUCACCGGCGCUGCACACAGUGACGCCCUUGCAUUCUCUGGCCCGAUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012180
          miRBase ID: bdi-miR397a
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Camelina sativa

          Stem-loop ID cas-MIR397
          Stem-loop Accession # MI0037226
          Stem-loop Sequence
          AACAUCAUUGAGUGCAGCGUUGAUGUGAUUUCGUUUUUUCAUUCUUUCAUUCUAUUAUUGAAUGGAUUGGAUUAAAGCAAUUUACACCAGCGUUGCGCUCAAUUAUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045348
          miRBase ID: cas-miR397
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Citrus sinensis

          Stem-loop ID csi-MIR397
          Stem-loop Accession # MI0016708
          Stem-loop Sequence
          CCAGAUUGAAAAAAACAUCAUUGAGUGCAGCGUUGAUGAAUAUUUGUCGGUGCUUUGCCAAUAUUUCUCUAUACUAGCCAGGAUUCUUUCACCAGCGCUGCACUCGAUCAUGUUUUUUAGCUCUGCUGGUUCAGGAUUAUCCAGAUAAUACGCACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018465
          miRBase ID: csi-miR397-5p
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Cucumis melo

          Stem-loop ID cme-MIR397
          Stem-loop Accession # MI0023252
          Stem-loop Sequence
          GAAAGAUACGUCAUUGAGUGCAGCGUUGAUGUCGAAACUUAUUUCAAAGUUCUAACUUUUCUCAUCAGCGCUUCAUUCAAUCACGUUUCUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026148
          miRBase ID: cme-miR397
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Fragaria vesca

          Stem-loop ID fve-MIR397
          Stem-loop Accession # MI0036414
          Stem-loop Sequence
          UUCAGAAUGAAGAAACGUCAUUGAGUGCAGCGUUGAUGAAGUCCUUAGUUUCGUUUCCGAGUUCCGAUGAUCUUCAUGGAGGCGAGCUGGAGGCAAUUUCAUCAACGCUGCACCCAAUUAUGUUUCUAUUCCUUCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044552
          miRBase ID: fve-miR397
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Glycine max

          Stem-loop ID gma-MIR397a
          Stem-loop Accession # MI0018645
          Stem-loop Sequence
          AGAGAAACAUCAUUGAGUGCAGCGUUGAUGAAGUUUCACUCUCAUCUCAGGUAGAUGCUUAAUUUAUAGUGUUAUUGUCAUCGACACUGCACUCAAUCAUGUUUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021627
          miRBase ID: gma-miR397a
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Stem-loop ID gma-MIR397b
          Stem-loop Accession # MI0018646
          Stem-loop Sequence
          GGAGAAACAUCAUUGAGUGCAGCGUUGAUGAAGUCCUAAAUAGUAAAUAGCAACUACCCCUCGUGUUUCUCAGGUAGAUGCUUAAAUGGUUUUAUUGACGCUGCACUCAAUCAUGUUUUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021628
          miRBase ID: gma-miR397b-5p
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Linum usitatissimum

          Stem-loop ID lus-MIR397b
          Stem-loop Accession # MI0021201
          Stem-loop Sequence
          GAGAAACCUCAUUGAGUGCAGCGUUGAUGAAAUUCAUCCCUUUUGCAGUCGGGAGGAUCUUCAGCAAUGCUGCUCUCAAUAACGCUUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0027181
          miRBase ID: lus-miR397b
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Manihot esculenta

          Stem-loop ID mes-MIR397b
          Stem-loop Accession # MI0037906
          Stem-loop Sequence
          AAUAAGAUUCCAGAUUGGAGAAACAUCAUUGAGUGCAGCGUUGAUGAAUUCCCUGCAAUUUUCUGGAAUUUGGGGAUGAUUUCACCAGCGCUGCAUUCAAUCAUGUUUUUUCACUUACUGGAAACGUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045970
          miRBase ID: mes-miR397b
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Medicago truncatula

          Stem-loop ID mtr-MIR397
          Stem-loop Accession # MI0025221
          Stem-loop Sequence
          UUGGAGAAACAUCAUUGAGUGCAGCGUUGAUGAAAUCUUAUGUAGCAUCUAGCUAUCUUAUAGAUAUCAAAUAUUGCAUGCUUAAUUUCAUCUACGCUACACUCAAUUAUGUUUUUUUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029984
          miRBase ID: mtr-miR397-5p
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Populus trichocarpa

          Stem-loop ID ptc-MIR397a
          Stem-loop Accession # MI0002332
          Stem-loop Sequence
          UGGAGAACCAUCAUUGAGUGCAGCGUUGAUGAAAUCCUCCAUUUUGUGCUAUUAAACUGUUACCAACCCUUUAUGGGGCAUGGCAUCAUUUCACCAGCGCUGCAUUCAAUCAUGUUUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002038
          miRBase ID: ptc-miR397a
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Prunus persica

          Stem-loop ID ppe-MIR397
          Stem-loop Accession # MI0026120
          Stem-loop Sequence
          AAGGGUGGCUCAAAAUCAAUCAUGCAGAUGCAGAUUAUGAGAGCAGUGGAGGUUCCAGAUUGAAGAAACAUCAUUGAGUGCAGCGUUGAUGAAUUGAAAUUCUUUCGAAAUUUCAUCAGCGCUGCACCCAAUUAUGUUUCUAUCCAAUUUUGGCAAAGUACUGGCUGGCUUCUCUGGUCUUCUUCUUUCUUUUAGCCCUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031496
          miRBase ID: ppe-miR397
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Rice

          Stem-loop ID osa-MIR397a
          Stem-loop Accession # MI0001049
          Stem-loop Sequence
          AUCAAAUGCAUCAUUGAGUGCAGCGUUGAUGAACAACGGUAACCGGUCCAUGUUGAUGCGCAUUUGGCCGGUGAUCUGAUCAUCAUCAGCGCUUCACUCAAUCAUGCGUUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000980
          miRBase ID: osa-miR397a
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Ricinus communis

          Stem-loop ID rco-MIR397
          Stem-loop Accession # MI0013419
          Stem-loop Sequence
          AAACAUCAUUGAGUGCAGCGUUGAUGAAUUCCCCUACAUGACAUAUUUCUGUUGGUUAAUUCAUUACCAAUAUCCCUUCUUUAAAAAAAGAAGUAAACCUUUUUGGGAAUAGAUGGUUUUGGAUGAUUUCACCAGCGCUGCAUUCAAUCAUGUUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014193
          miRBase ID: rco-miR397
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Salvia sclarea

          Stem-loop ID ssl-MIR397
          Stem-loop Accession # MI0019352
          Stem-loop Sequence
          UCAUUGAGUGCAGCGUUGAUGACAUCGGCAUUCGUCCUUUUCCUUCGUCUCCGACUGUCGUUUCAUUGACGCUGCACUCAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022518
          miRBase ID: ssl-miR397
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Sorghum bicolor

          Stem-loop ID sbi-MIR397
          Stem-loop Accession # MI0010899
          Stem-loop Sequence
          UCAUUGAGUGCAGCGUUGAUGAGCCAGCUGGCCGGCCAGCCGUGCGUCCGCCGCCGGCGCCGGCCACGGCUCACCGGCGCUGCACUCAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011359
          miRBase ID: sbi-miR397-5p
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Theobroma cacao

          Stem-loop ID tcc-MIR397
          Stem-loop Accession # MI0017516
          Stem-loop Sequence
          AUUCCCAGAUGGAAGACACAUCAUUGAGUGCAGCGUUGAUGAAUUCUUUCACUUUUUGCCAACUCUAUCUCAGAUGAAUUCGCCAGCGCUGCACUCAAUCAUGUGUUUUGCUCUUUCUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020432
          miRBase ID: tcc-miR397
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Vitis vinifera

          Stem-loop ID vvi-MIR397a
          Stem-loop Accession # MI0007956
          Stem-loop Sequence
          GAAGAAAACAUCAUUGAGUGCAGCGUUGAUGAAACUGAAGUAUUCCAUUUUUCAGCUUCUUUGAAGUCCGGCAAGAUGGGUUCUGGUUGAUUCCAUUGGCGCUGCACUCAAUCAUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006561
          miRBase ID: vvi-miR397a
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA

          Vriesea carinata

          Stem-loop ID vca-MIR397
          Stem-loop Accession # MI0032994
          Stem-loop Sequence
          GCAUCAUUGAGUGCAGCGUUGAUGACCGUCGGGAACGCAUAGUUUGAGAGGUUUCAUCAGCGCUUCACUCAAUCAUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041015
          miRBase ID: vca-miR397-5p
          Mature miRNA Sequence: UCAUUGAGUGCAGCGUUGAUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 000368
          mirVana® miRNA inhibitor : MH10836


          miRBase Alias:

          Arabidopsis lyrata: aly-miR397a (v18)
          Arabidopsis lyrata: aly-miR397b (v18)
          Citrus sinensis: csi-miR397 (v21)
          Glycine max: gma-miR397b (v18)
          Shewanella bicestrii: sbi-miR397 (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline