Search
Search

| Assay Name: | ath-miR397a |
|---|---|
| miRBase Accession Number: | MI0001015 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAUUGAGUGCAGCGUUGAUG |
| Species: | Arabidopsis, Arabidopsis lyrata, Brachypodium distachyon, Camelina sativa, Citrus sinensis, Cucumis melo, Fragaria vesca, Glycine max, Linum usitatissimum, Manihot esculenta, Medicago truncatula, Populus trichocarpa, Prunus persica, Rice, Ricinus communis, Salvia sclarea, Sorghum bicolor, Theobroma cacao, Vitis vinifera, Vriesea carinata |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000120, MIR397 |
Mature miRNA Sequence: | UCAUUGAGUGCAGCGUUGAUG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Arabidopsis | ath-miR397a | MIMAT0000946 | |
| Arabidopsis lyrata | aly-miR397a-5p | MIMAT0017561 | |
| Arabidopsis lyrata | aly-miR397b-5p | MIMAT0017563 | |
| Brachypodium distachyon | bdi-miR397a | MIMAT0012180 | |
| Camelina sativa | cas-miR397 | MIMAT0045348 | |
| Citrus sinensis | csi-miR397-5p | MIMAT0018465 | |
| View More Rows | |||
| Cucumis melo | cme-miR397 | MIMAT0026148 | |
| Fragaria vesca | fve-miR397 | MIMAT0044552 | |
| Glycine max | gma-miR397a | MIMAT0021627 | |
| Glycine max | gma-miR397b-5p | MIMAT0021628 | |
| Linum usitatissimum | lus-miR397b | MIMAT0027181 | |
| Manihot esculenta | mes-miR397b | MIMAT0045970 | |
| Medicago truncatula | mtr-miR397-5p | MIMAT0029984 | |
| Populus trichocarpa | ptc-miR397a | MIMAT0002038 | |
| Prunus persica | ppe-miR397 | MIMAT0031496 | |
| Rice | osa-miR397a | MIMAT0000980 | |
| Ricinus communis | rco-miR397 | MIMAT0014193 | |
| Salvia sclarea | ssl-miR397 | MIMAT0022518 | |
| Sorghum bicolor | sbi-miR397-5p | MIMAT0011359 | |
| Theobroma cacao | tcc-miR397 | MIMAT0020432 | |
| Vitis vinifera | vvi-miR397a | MIMAT0006561 | |
| Vriesea carinata | vca-miR397-5p | MIMAT0041015 | |