Search
Search

| Assay Name: | cgr-miR-149-5p |
|---|---|
| miRBase Accession Number: | MI0020411 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUGGCUCCGUGUCUUCACUCC |
| Species: | Cricetulus griseus, Nomascus leucogenys, Otolemur garnettii |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000274, mir-149 |
Mature miRNA Sequence: | UCUGGCUCCGUGUCUUCACUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-149-5p | MIMAT0023780 |
| Nomascus leucogenys | nle-miR-149 | MIMAT0049345 |
| Otolemur garnettii | oga-miR-149 | MIMAT0049706 |