Search
Search

| Assay Name: | dre-miR-199-3p |
|---|---|
| miRBase Accession Number: | MI0001373 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACAGUAGUCUGCACAUUGGUU |
| Species: | Ophiophagus hannah, Pig, Xenopus tropicalis, Zebrafish |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000040, mir-199 |
Mature miRNA Sequence: | UACAGUAGUCUGCACAUUGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ophiophagus hannah | oha-miR-199c-3p | MIMAT0036794 |
| Pig | ssc-miR-199b-3p | MIMAT0007760 |
| Xenopus tropicalis | xtr-miR-199a-3p | MIMAT0003619 |
| Zebrafish | dre-miR-199-3p | MIMAT0003155 |