Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH11072
          Assay Name: dre-miR-199-3p
          miRBase Accession Number: MI0001373
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UACAGUAGUCUGCACAUUGGUU
          Species: Ophiophagus hannah, Pig, Xenopus tropicalis, Zebrafish
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH11072

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000040, mir-199

          Mature miRNA Sequence:

          UACAGUAGUCUGCACAUUGGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Ophiophagus hannah oha-miR-199c-3p MIMAT0036794
          Pig ssc-miR-199b-3p MIMAT0007760
          Xenopus tropicalis xtr-miR-199a-3p MIMAT0003619
          Zebrafish dre-miR-199-3p MIMAT0003155

          Stem-loop Details



          Ophiophagus hannah

          Stem-loop ID oha-mir-199c
          Stem-loop Accession # MI0031394
          Stem-loop Sequence
          UCAACCUGCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCUGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCAAGGGAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036794
          miRBase ID: oha-miR-199c-3p
          Mature miRNA Sequence: UACAGUAGUCUGCACAUUGGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-199b
          Stem-loop Accession # MI0008219
          Stem-loop Sequence
          CCGUCUACCCAGUGUUUAGACUAUCUGUUCAGGACUCCCAAAUUGUACAGUAGUCUGCACAUUGGUUAGGCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007760
          miRBase ID: ssc-miR-199b-3p
          Mature miRNA Sequence: UACAGUAGUCUGCACAUUGGUU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-199a
          Stem-loop Accession # MI0004859
          Stem-loop Sequence
          AAGAACCUGCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCUGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCAUGGGACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003619
          miRBase ID: xtr-miR-199a-3p
          Mature miRNA Sequence: UACAGUAGUCUGCACAUUGGUU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-199-1
          Stem-loop Accession # MI0001373
          Stem-loop Sequence
          UCCUGCUCCGUCAUCCCAGUGUUCAGACUACCUGUUCAGGAUCAUACUGGUGUACAGUAGUCUGCACAUUGGUUAGACUGUGCAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003155
          miRBase ID: dre-miR-199-3p
          Mature miRNA Sequence: UACAGUAGUCUGCACAUUGGUU
          Chromosome Location: NA

          Stem-loop ID dre-mir-199-2
          Stem-loop Accession # MI0001374
          Stem-loop Sequence
          GGAGUUUUUGUGGACGCCCGUCCCGCCUGCCCAGUGUUCAGACUACCUGUUCAGGAAUUAGUGUUUGUACAGUAGUCUGCACAUUGGUUAGGCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003155
          miRBase ID: dre-miR-199-3p
          Mature miRNA Sequence: UACAGUAGUCUGCACAUUGGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11072
          Ambion® Pre-miR™ miRNA Precursor : PM11072
          TaqMan™ MicroRNA Assay : 000499
          mirVana® miRNA mimic : MC11072


          miRBase Alias:

          Danio rerio: dre-miR-199* (v19)
          Staphylococcus schleiferi: ssc-miR-199b* (v18)
          Xenopus tropicalis: xtr-miR-199a* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline