Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11157
          Assay Name: rno-miR-125b-2-3p
          miRBase Accession Number: MI0000897
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 16097346 - 16097433 [+] on Build Rnor_6.0
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Species: Rat, Chicken, Chrysemys picta, Columba livia, Dasypus novemcinctus, Guinea pig, Python bivittatus, Rabbit, Rhesus monkey, Taeniopygia guttata
          Product Type: mirVana® miRNA mimic
          Assay ID MC11157

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          ACAAGUCAGGCUCUUGGGACCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-125b-2-3p MIMAT0026467
          Chicken gga-miR-125b-3p MIMAT0026493
          Chrysemys picta cpi-miR-125b-2-3p MIMAT0037732
          Columba livia cli-miR-125-2-3p MIMAT0038498
          Dasypus novemcinctus dno-miR-125b-2-3p MIMAT0047616
          Guinea pig cpo-miR-125b-2-3p MIMAT0046969
          View More Rows
          Python bivittatus pbv-miR-125b-3p MIMAT0038917
          Rabbit ocu-miR-125b-2-3p MIMAT0048207
          Rhesus monkey mml-miR-125b-2-3p MIMAT0031073
          Taeniopygia guttata tgu-miR-125-1-3p MIMAT0014628

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-125b-2
          Stem-loop Accession # MI0000897
          Stem-loop Sequence
          ACCAGACUUUUCCUAGUCCCUGAGACCCUAACUUGUGAGGUAUUUUAGUAACAUCACAAGUCAGGCUCUUGGGACCUAGGCGGAGAGG
          Chromosome Location Chr. 11 - 16097346 - 16097433 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0026467
          miRBase ID: rno-miR-125b-2-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: Chr. 11 - 16097346 - 16097433 [+] on Build Rnor_6.0

          Chicken

          Stem-loop ID gga-mir-125b-2
          Stem-loop Accession # MI0001175
          Stem-loop Sequence
          AUCAGACUUUUCCUAGUCCCUGAGACCCUAACUUGUGAGGUUUUGUAGCAACAAUCACAAGUCAGGCUCUUGGGACCUAGGCGGAGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026493
          miRBase ID: gga-miR-125b-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-125b-2
          Stem-loop Accession # MI0029461
          Stem-loop Sequence
          CGGACUUUUCCUAGUCCCUGAGACCCUAACUUGUGAGGUUUUUUAGCAACAAUCACAAGUCAGGCUCUUGGGACCUAGGCGGAGGGGAACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037732
          miRBase ID: cpi-miR-125b-2-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-125-2
          Stem-loop Accession # MI0029957
          Stem-loop Sequence
          GGACUUUUCCUAGUCCCUGAGACCCUAACUUGUGAGGUUUUUUAGCAACAAUCACAAGUCAGGCUCUUGGGACCUAGGCGGAGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038498
          miRBase ID: cli-miR-125-2-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-125b-2
          Stem-loop Accession # MI0038967
          Stem-loop Sequence
          UCCCUGAGACCCUAACUUGUGAGGUAUUUUAGUAACAUCACAAGUCAGGCUCUUGGGACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047616
          miRBase ID: dno-miR-125b-2-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-125b-2
          Stem-loop Accession # MI0038626
          Stem-loop Sequence
          UCCCUGAGACCCUAACUUGUGAGGUAUUUAGUAACAUCACAAGUCAGGCUCUUGGGACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046969
          miRBase ID: cpo-miR-125b-2-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-125b-2
          Stem-loop Accession # MI0030217
          Stem-loop Sequence
          GACUUUUCCUAGUCCCUGAGACCCUAACUUGUGAGGUUUUUUAGUAACAAUCACAAGUCAGGCUCUUGGGACCUAGGCGGAGGGGAACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038917
          miRBase ID: pbv-miR-125b-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-125b-2
          Stem-loop Accession # MI0039279
          Stem-loop Sequence
          UCCCUGAGACCCUAACUUGUGAGGUAUUUUAGUAACAUCACAAGUCAGGCUCUUGGGACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048207
          miRBase ID: ocu-miR-125b-2-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-125b-2
          Stem-loop Accession # MI0002579
          Stem-loop Sequence
          ACCAGACUUUUCCUAGUCCCUGAGACCCUAACUUGUGAGGUAUUUUAGUAACAUCACAAGUCAGGCUCUUGGGACCUAGGCGGAGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031073
          miRBase ID: mml-miR-125b-2-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-125-1
          Stem-loop Accession # MI0013695
          Stem-loop Sequence
          CUUUUCCUAGUCCCUGAGACCCUAACUUGUGAGGUUUUUUAGCAACAAUCACAAGUCAGGCUCUUGGGACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014628
          miRBase ID: tgu-miR-125-1-3p
          Mature miRNA Sequence: ACAAGUCAGGCUCUUGGGACCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465041_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11157
          Ambion® Pre-miR™ miRNA Precursor : PM11157
          TaqMan™ MicroRNA Assay : 002074
          mirVana® miRNA inhibitor : MH11157
          TaqMan™ Advanced miRNA Assay : rno480879_mir


          miRBase Alias:

          Rat: rno-miR-125b* (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-125-1* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline