Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11407
          Assay Name: ggo-miR-615
          miRBase Accession Number: MI0039831
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCU
          Species: Gorilla gorilla, Microcebus murinus, Otolemur garnettii, Pan paniscus, Pig
          Product Type: mirVana® miRNA mimic
          Assay ID MC11407

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Mature miRNA Sequence:

          UCCGAGCCUGGGUCUCCCUCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Gorilla gorilla ggo-miR-615 MIMAT0049119
          Microcebus murinus mmr-miR-615 MIMAT0049125
          Otolemur garnettii oga-miR-615 MIMAT0049637
          Pan paniscus ppa-miR-615 MIMAT0049056
          Pig ssc-miR-615 MIMAT0020595

          Stem-loop Details



          Gorilla gorilla

          Stem-loop ID ggo-mir-615
          Stem-loop Accession # MI0039831
          Stem-loop Sequence
          GGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049119
          miRBase ID: ggo-miR-615
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-615
          Stem-loop Accession # MI0039837
          Stem-loop Sequence
          GGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049125
          miRBase ID: mmr-miR-615
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-615
          Stem-loop Accession # MI0040420
          Stem-loop Sequence
          AGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049637
          miRBase ID: oga-miR-615
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-615
          Stem-loop Accession # MI0039764
          Stem-loop Sequence
          GGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049056
          miRBase ID: ppa-miR-615
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-615
          Stem-loop Accession # MI0017995
          Stem-loop Sequence
          GGGAGGGGCGGGAGGGGGCUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUCCCCCCCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020595
          miRBase ID: ssc-miR-615
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11407
          Ambion® Pre-miR™ miRNA Precursor : PM11407
          TaqMan™ MicroRNA Assay : 001588
          mirVana® miRNA inhibitor : MH11407

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline