Search
Search

| Assay Name: | |
|---|---|
| miRBase Accession Number: | MI0039831 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCGAGCCUGGGUCUCCCUCU |
| Species: | Gorilla gorilla, Microcebus murinus, Otolemur garnettii, Pan paniscus, Pig |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UCCGAGCCUGGGUCUCCCUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-615 | MIMAT0049119 |
| Microcebus murinus | mmr-miR-615 | MIMAT0049125 |
| Otolemur garnettii | oga-miR-615 | MIMAT0049637 |
| Pan paniscus | ppa-miR-615 | MIMAT0049056 |
| Pig | ssc-miR-615 | MIMAT0020595 |