Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11758
          Assay Name: hsa-let-7g-5p
          miRBase Accession Number: MI0000433
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 52268278 - 52268361 [-] on Build GRCh38
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Bovine, Callithrix jacchus, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Ophiophagus hannah, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Taeniopygia guttata, Tupaia chinensis
          Product Type: mirVana® miRNA mimic
          Assay ID MC11758

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          UGAGGUAGUAGUUUGUACAGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7g-5p MIMAT0000414
          Mouse mmu-let-7g-5p MIMAT0000121
          Rat rno-let-7g-5p MIMAT0035719
          Alligator mississippiensis ami-let-7g-5p MIMAT0038014
          Anolis carolinensis aca-let-7g MIMAT0021706
          Bovine bta-let-7g MIMAT0003838
          View More Rows
          Callithrix jacchus cja-let-7g MIMAT0039326
          Chrysemys picta cpi-let-7g-5p MIMAT0037608
          Columba livia cli-let-7g-5p MIMAT0038387
          Cricetulus griseus cgr-let-7g-5p MIMAT0023722
          Dasypus novemcinctus dno-let-7g-5p MIMAT0047485
          Daubentonia madagascariensis dma-let-7g MIMAT0049290
          Dog cfa-let-7g MIMAT0006637
          Goat chi-let-7g-5p MIMAT0035892
          Guinea pig cpo-let-7g-5p MIMAT0046831
          Horse eca-let-7g MIMAT0013075
          Microcebus murinus mmr-let-7g MIMAT0049151
          Nomascus leucogenys nle-let-7g MIMAT0049456
          Ophiophagus hannah oha-let-7g-5p MIMAT0036640
          Ornithorhynchus anatinus oan-let-7g-5p MIMAT0007038
          Otolemur garnettii oga-let-7g MIMAT0049649
          Pan paniscus ppa-let-7g MIMAT0049075
          Pan troglodytes ptr-let-7g MIMAT0007942
          Papio hamadryas pha-let-7g MIMAT0049574
          Pig ssc-let-7g MIMAT0013867
          Pongo pygmaeus ppy-let-7g MIMAT0015727
          Pteropus alecto pal-let-7g-5p MIMAT0039774
          Python bivittatus pbv-let-7g-5p MIMAT0038808
          Rabbit ocu-let-7g-5p MIMAT0048087
          Rhesus monkey mml-let-7g-5p MIMAT0006157
          Taeniopygia guttata tgu-let-7g-5p MIMAT0014528
          Tupaia chinensis tch-let-7g-5p MIMAT0036519

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7g
          Stem-loop Accession # MI0000433
          Stem-loop Sequence
          AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCA
          Chromosome Location Chr. 3 - 52268278 - 52268361 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000414
          miRBase ID: hsa-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: Chr. 3 - 52268278 - 52268361 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7g
          Stem-loop Accession # MI0000137
          Stem-loop Sequence
          CCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGG
          Chromosome Location Chr. 9 - 106178840 - 106178927 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000121
          miRBase ID: mmu-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: Chr. 9 - 106178840 - 106178927 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7g
          Stem-loop Accession # MI0030347
          Stem-loop Sequence
          CCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGG
          Chromosome Location Chr. 8 - 114874457 - 114874544 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0035719
          miRBase ID: rno-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: Chr. 8 - 114874457 - 114874544 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-let-7g
          Stem-loop Accession # MI0029650
          Stem-loop Sequence
          GCCUGAUUCCAAGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUUAACUGUACAGGCCACUGCCUUGCCUGGGACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038014
          miRBase ID: ami-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-let-7g
          Stem-loop Accession # MI0018709
          Stem-loop Sequence
          UUGCCUGAUUCCGGGUUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGGCAGAGCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021706
          miRBase ID: aca-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-let-7g
          Stem-loop Accession # MI0005051
          Stem-loop Sequence
          AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003838
          miRBase ID: bta-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-let-7g
          Stem-loop Accession # MI0031835
          Stem-loop Sequence
          GCUCCGUUUCCUUUUGCCUGAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGGAACAGUGCGCCAGCUGCCAAAUGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039326
          miRBase ID: cja-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7g
          Stem-loop Accession # MI0029382
          Stem-loop Sequence
          UCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037608
          miRBase ID: cpi-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-let-7g
          Stem-loop Accession # MI0029891
          Stem-loop Sequence
          UUCUUGCCUGAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038387
          miRBase ID: cli-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-let-7g
          Stem-loop Accession # MI0020373
          Stem-loop Sequence
          AUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGGAACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023722
          miRBase ID: cgr-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7g
          Stem-loop Accession # MI0038893
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047485
          miRBase ID: dno-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-let-7g
          Stem-loop Accession # MI0040023
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049290
          miRBase ID: dma-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-let-7g
          Stem-loop Accession # MI0008036
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006637
          miRBase ID: cfa-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7g
          Stem-loop Accession # MI0030585
          Stem-loop Sequence
          CCUUUUGCCUGAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCGGGAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035892
          miRBase ID: chi-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7g
          Stem-loop Accession # MI0038549
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUAUAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046831
          miRBase ID: cpo-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-let-7g
          Stem-loop Accession # MI0012820
          Stem-loop Sequence
          CCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013075
          miRBase ID: eca-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-let-7g
          Stem-loop Accession # MI0039866
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUAUAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049151
          miRBase ID: mmr-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-let-7g
          Stem-loop Accession # MI0040208
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049456
          miRBase ID: nle-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-let-7g
          Stem-loop Accession # MI0031309
          Stem-loop Sequence
          GGUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036640
          miRBase ID: oha-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-let-7g
          Stem-loop Accession # MI0006809
          Stem-loop Sequence
          UCCAGGCUGAGGUAGUAGUUUGUACAGUUUCAGGGUCUACAAUACCACCCGGUAUAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007038
          miRBase ID: oan-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-let-7g
          Stem-loop Accession # MI0040432
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUGUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049649
          miRBase ID: oga-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-let-7g
          Stem-loop Accession # MI0039783
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049075
          miRBase ID: ppa-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-let-7g
          Stem-loop Accession # MI0008406
          Stem-loop Sequence
          AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007942
          miRBase ID: ptr-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-let-7g
          Stem-loop Accession # MI0040349
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049574
          miRBase ID: pha-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-let-7g
          Stem-loop Accession # MI0013087
          Stem-loop Sequence
          GGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013867
          miRBase ID: ssc-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-let-7g
          Stem-loop Accession # MI0014788
          Stem-loop Sequence
          AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015727
          miRBase ID: ppy-let-7g
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-let-7g
          Stem-loop Accession # MI0032321
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039774
          miRBase ID: pal-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7g
          Stem-loop Accession # MI0030141
          Stem-loop Sequence
          GUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUACGAUACCGCCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038808
          miRBase ID: pbv-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-let-7g
          Stem-loop Accession # MI0039213
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048087
          miRBase ID: ocu-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7g
          Stem-loop Accession # MI0007579
          Stem-loop Sequence
          AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006157
          miRBase ID: mml-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7g-1
          Stem-loop Accession # MI0013740
          Stem-loop Sequence
          GAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014528
          miRBase ID: tgu-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Stem-loop ID tgu-let-7g-2
          Stem-loop Accession # MI0013741
          Stem-loop Sequence
          GAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014528
          miRBase ID: tgu-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-let-7g
          Stem-loop Accession # MI0031177
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036519
          miRBase ID: tch-let-7g-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUACAGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306155_pri
          TaqMan™ Pri-miRNA Assay : Hs03303029_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11758
          Ambion® Pre-miR™ miRNA Precursor : PM11758
          TaqMan™ MicroRNA Assay : 002282
          mirVana® miRNA inhibitor : MH11758
          TaqMan™ Advanced miRNA Assay : 478580_mir
          TaqMan™ Advanced miRNA Assay : rno478580_mir


          miRBase Alias:

          Human: hsa-let-7g (v17)
          Mouse: mmu-let-7g (v17)
          Macaca mulatta: mml-let-7g (v19)
          Ochrobactrum anthropi: oan-let-7g (v18)
          Trabulsiella guamensis ATCC 49490: tgu-let-7g (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline