Search
Search

| Assay Name: | rno-let-7g-5p |
|---|---|
| Stem-loop Accession Number: | MI0000433 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGGUAGUAGUUUGUACAGUU |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Bovine, Callithrix jacchus, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Ophiophagus hannah, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Taeniopygia guttata, Tupaia chinensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | UGAGGUAGUAGUUUGUACAGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7g-5p | MIMAT0000414 |
| Mouse | mmu-let-7g-5p | MIMAT0000121 |
| Rat | rno-let-7g-5p | MIMAT0035719 |
| Alligator mississippiensis | ami-let-7g-5p | MIMAT0038014 |
| Anolis carolinensis | aca-let-7g | MIMAT0021706 |
| Bovine | bta-let-7g | MIMAT0003838 |
| View More Rows | ||
| Callithrix jacchus | cja-let-7g | MIMAT0039326 |
| Chrysemys picta | cpi-let-7g-5p | MIMAT0037608 |
| Columba livia | cli-let-7g-5p | MIMAT0038387 |
| Cricetulus griseus | cgr-let-7g-5p | MIMAT0023722 |
| Dasypus novemcinctus | dno-let-7g-5p | MIMAT0047485 |
| Daubentonia madagascariensis | dma-let-7g | MIMAT0049290 |
| Dog | cfa-let-7g | MIMAT0006637 |
| Goat | chi-let-7g-5p | MIMAT0035892 |
| Guinea pig | cpo-let-7g-5p | MIMAT0046831 |
| Horse | eca-let-7g | MIMAT0013075 |
| Microcebus murinus | mmr-let-7g | MIMAT0049151 |
| Nomascus leucogenys | nle-let-7g | MIMAT0049456 |
| Ophiophagus hannah | oha-let-7g-5p | MIMAT0036640 |
| Ornithorhynchus anatinus | oan-let-7g-5p | MIMAT0007038 |
| Otolemur garnettii | oga-let-7g | MIMAT0049649 |
| Pan paniscus | ppa-let-7g | MIMAT0049075 |
| Pan troglodytes | ptr-let-7g | MIMAT0007942 |
| Papio hamadryas | pha-let-7g | MIMAT0049574 |
| Pig | ssc-let-7g | MIMAT0013867 |
| Pongo pygmaeus | ppy-let-7g | MIMAT0015727 |
| Pteropus alecto | pal-let-7g-5p | MIMAT0039774 |
| Python bivittatus | pbv-let-7g-5p | MIMAT0038808 |
| Rabbit | ocu-let-7g-5p | MIMAT0048087 |
| Rhesus monkey | mml-let-7g-5p | MIMAT0006157 |
| Taeniopygia guttata | tgu-let-7g-5p | MIMAT0014528 |
| Tupaia chinensis | tch-let-7g-5p | MIMAT0036519 |