Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11842
          Assay Name: hsa-miR-770-5p
          miRBase Accession Number: MI0005118
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100852390 - 100852487 [+] on Build GRCh38
          Mature miRNA Sequence: UCCAGUACCACGUGUCAGGGCCA
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC11842

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000355, mir-770

          Mature miRNA Sequence:

          UCCAGUACCACGUGUCAGGGCCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-770-5p MIMAT0003948
          Pan troglodytes ptr-miR-770 MIMAT0008322
          Pongo pygmaeus ppy-miR-770-5p MIMAT0016109
          Rhesus monkey mml-miR-770-5p MIMAT0006514

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-770
          Stem-loop Accession # MI0005118
          Stem-loop Sequence
          AGGAGCCACCUUCCGAGCCUCCAGUACCACGUGUCAGGGCCACAUGAGCUGGGCCUCGUGGGCCUGAUGUGGUGCUGGGGCCUCAGGGGUCUGCUCUU
          Chromosome Location Chr. 14 - 100852390 - 100852487 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003948
          miRBase ID: hsa-miR-770-5p
          Mature miRNA Sequence: UCCAGUACCACGUGUCAGGGCCA
          Chromosome Location: Chr. 14 - 100852390 - 100852487 [+] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-770
          Stem-loop Accession # MI0008861
          Stem-loop Sequence
          GGAGCCACCUUCCGAGCCUCCAGUACCACGUGUCAGGGCCACAUGAGCUGGGCCUCGUGGGCCUGAUGUGGUGCUGGGGCCUCAGGGGUCUGCUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008322
          miRBase ID: ptr-miR-770
          Mature miRNA Sequence: UCCAGUACCACGUGUCAGGGCCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-770
          Stem-loop Accession # MI0015148
          Stem-loop Sequence
          AGGAGCCACCUUCCGAGCCUCCAGUACCACGUGUCAGGGCCACAUGAGCUGGGCCUCGUGGGCCUGAUGUGGUGCUGGGGCCUCAGGGGUCUGCUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016109
          miRBase ID: ppy-miR-770-5p
          Mature miRNA Sequence: UCCAGUACCACGUGUCAGGGCCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-770
          Stem-loop Accession # MI0007913
          Stem-loop Sequence
          AGGAGCCACCUUCCGAGCCUCCAGUACCACGUGUCAGGGCCACAUGAGCUGGGCCUCGUGGGCCUGAUGUGGUGCUGGGGCCUCAGGGGUCUGCUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006514
          miRBase ID: mml-miR-770-5p
          Mature miRNA Sequence: UCCAGUACCACGUGUCAGGGCCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305042_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11842
          Ambion® Pre-miR™ miRNA Precursor : PM11842
          TaqMan™ MicroRNA Assay : 002002
          mirVana® miRNA inhibitor : MH11842
          TaqMan™ Advanced miRNA Assay : 479178_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0