Search
Search

| Assay Name: | hsa-miR-770-5p |
|---|---|
| miRBase Accession Number: | MI0005118 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCAGUACCACGUGUCAGGGCCA |
| Species: | Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000355, mir-770 |
Mature miRNA Sequence: | UCCAGUACCACGUGUCAGGGCCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-770-5p | MIMAT0003948 |
| Pan troglodytes | ptr-miR-770 | MIMAT0008322 |
| Pongo pygmaeus | ppy-miR-770-5p | MIMAT0016109 |
| Rhesus monkey | mml-miR-770-5p | MIMAT0006514 |