Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12369
          Assay Name: hsa-miR-182-5p
          miRBase Accession Number: MI0000272
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 129770383 - 129770492 [-] on Build GRCh38
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Species: Human, Alligator mississippiensis, Bovine, Chrysemys picta, Columba livia, Dasypus novemcinctus, Dog, Guinea pig, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan troglodytes, Petromyzon marinus, Pig, Python bivittatus, Rabbit
          Product Type: mirVana® miRNA mimic
          Assay ID MC12369

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000116, mir-182

          Mature miRNA Sequence:

          UUUGGCAAUGGUAGAACUCACACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-182-5p MIMAT0000259
          Alligator mississippiensis ami-miR-182-5p MIMAT0038194
          Bovine bta-miR-182 MIMAT0009244
          Chrysemys picta cpi-miR-182-5p MIMAT0037798
          Columba livia cli-miR-182-5p MIMAT0038556
          Dasypus novemcinctus dno-miR-182-5p MIMAT0047699
          View More Rows
          Dog cfa-miR-182 MIMAT0009841
          Guinea pig cpo-miR-182-5p MIMAT0047048
          Microcebus murinus mmr-miR-182 MIMAT0049216
          Nomascus leucogenys nle-miR-182 MIMAT0049442
          Otolemur garnettii oga-miR-182 MIMAT0049650
          Pan troglodytes ptr-miR-182 MIMAT0008050
          Petromyzon marinus pma-miR-182 MIMAT0019490
          Pig ssc-miR-182 MIMAT0025366
          Python bivittatus pbv-miR-182-5p MIMAT0038980
          Rabbit ocu-miR-182-5p MIMAT0048285

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-182
          Stem-loop Accession # MI0000272
          Stem-loop Sequence
          GAGCUGCUUGCCUCCCCCCGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAACAGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGGGCGAGGACUCAGCCGGCAC
          Chromosome Location Chr. 7 - 129770383 - 129770492 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000259
          miRBase ID: hsa-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: Chr. 7 - 129770383 - 129770492 [-] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-182
          Stem-loop Accession # MI0029767
          Stem-loop Sequence
          GGUCCCUCUCGCUGUCUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAUCAGGAUCCGGUGGUUCUAGACUUGCCAACUACCGCCCGAGGGCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038194
          miRBase ID: ami-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-182
          Stem-loop Accession # MI0009755
          Stem-loop Sequence
          CGCCGCCGCCUGCCUCCCACCAUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGUGCGAGGGCUUGGCCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009244
          miRBase ID: bta-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-182
          Stem-loop Accession # MI0029506
          Stem-loop Sequence
          UCUCUCUCGCUGUCUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAUCAGGAUCCGGUGGUUCUAGACUUGCCAACUACCGCCCGAGGGUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037798
          miRBase ID: cpi-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-182
          Stem-loop Accession # MI0029993
          Stem-loop Sequence
          CGGCCCCGGGGCUGCUUUUGGCAAUGGUAGAACUCACACUGGUGCGGUAGCAGGAUCCGGUGGUUCUAGACUUGCCAACUACAGCCCCGGGGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038556
          miRBase ID: cli-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-182
          Stem-loop Accession # MI0039013
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047699
          miRBase ID: dno-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-182
          Stem-loop Accession # MI0010336
          Stem-loop Sequence
          GCUUGCCUCCCGCCGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGUGCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009841
          miRBase ID: cfa-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-182
          Stem-loop Accession # MI0038670
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047048
          miRBase ID: cpo-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-182
          Stem-loop Accession # MI0039942
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUCAUGGGAUCCGGUGGUUCUAGACUUGCCAACUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049216
          miRBase ID: mmr-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-182
          Stem-loop Accession # MI0040189
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAACAGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049442
          miRBase ID: nle-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-182
          Stem-loop Accession # MI0040433
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049650
          miRBase ID: oga-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-182
          Stem-loop Accession # MI0008558
          Stem-loop Sequence
          GAGCUGCUUGCCUCCCUCCGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAACAGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGGGCGAGGACUCAGCCGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008050
          miRBase ID: ptr-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Petromyzon marinus

          Stem-loop ID pma-mir-182
          Stem-loop Accession # MI0017091
          Stem-loop Sequence
          GGCGGACUGUGUUUGGCAAUGGUAGAACUCACACUGGCCAGAGAGCUCUCCGGGGGUUCUAGACUUGCCAAUUACAGUCCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019490
          miRBase ID: pma-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-182
          Stem-loop Accession # MI0022135
          Stem-loop Sequence
          UCGCACCGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGAUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGUGCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025366
          miRBase ID: ssc-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-182
          Stem-loop Accession # MI0030259
          Stem-loop Sequence
          CUCUCCCUCUUGCUGUCUUUGGCAAUGGUAGAACUCACACUGGUGAGAUAUCUGGAUCCGGUGGUUCUAGACUUGCCAACUACCGCCUGGGGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038980
          miRBase ID: pbv-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-182
          Stem-loop Accession # MI0039321
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAUGGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048285
          miRBase ID: ocu-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302897_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12369
          Ambion® Pre-miR™ miRNA Precursor : PM12369
          TaqMan™ MicroRNA Assay : 002334
          mirVana® miRNA inhibitor : MH12369
          TaqMan™ Advanced miRNA Assay : 477935_mir


          miRBase Alias:

          Human: hsa-miR-182 (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline