Search
Search

| Assay Name: | hsa-miR-182-5p |
|---|---|
| miRBase Accession Number: | MI0000272 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUGGCAAUGGUAGAACUCACACU |
| Species: | Human, Alligator mississippiensis, Bovine, Chrysemys picta, Columba livia, Dasypus novemcinctus, Dog, Guinea pig, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan troglodytes, Petromyzon marinus, Pig, Python bivittatus, Rabbit |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000116, mir-182 |
Mature miRNA Sequence: | UUUGGCAAUGGUAGAACUCACACU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-182-5p | MIMAT0000259 |
| Alligator mississippiensis | ami-miR-182-5p | MIMAT0038194 |
| Bovine | bta-miR-182 | MIMAT0009244 |
| Chrysemys picta | cpi-miR-182-5p | MIMAT0037798 |
| Columba livia | cli-miR-182-5p | MIMAT0038556 |
| Dasypus novemcinctus | dno-miR-182-5p | MIMAT0047699 |
| View More Rows | ||
| Dog | cfa-miR-182 | MIMAT0009841 |
| Guinea pig | cpo-miR-182-5p | MIMAT0047048 |
| Microcebus murinus | mmr-miR-182 | MIMAT0049216 |
| Nomascus leucogenys | nle-miR-182 | MIMAT0049442 |
| Otolemur garnettii | oga-miR-182 | MIMAT0049650 |
| Pan troglodytes | ptr-miR-182 | MIMAT0008050 |
| Petromyzon marinus | pma-miR-182 | MIMAT0019490 |
| Pig | ssc-miR-182 | MIMAT0025366 |
| Python bivittatus | pbv-miR-182-5p | MIMAT0038980 |
| Rabbit | ocu-miR-182-5p | MIMAT0048285 |