Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12531
          Assay Name: hsa-miR-499a-3p
          miRBase Accession Number: MI0003183
          miRBase Version: v22.1
          Chromosome Location: Chr.20: 34990376 - 34990497 [+] on Build GRCh38
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Species: Human, Rat, Dasypus novemcinctus, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12531

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000173, mir-499

          Mature miRNA Sequence:

          AACAUCACAGCAAGUCUGUGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-499a-3p MIMAT0004772
          Rat rno-miR-499-3p MIMAT0017227
          Dasypus novemcinctus dno-miR-499-3p MIMAT0047937
          Guinea pig cpo-miR-499-3p MIMAT0047298
          Horse eca-miR-499-3p MIMAT0013109
          Pan troglodytes ptr-miR-499 MIMAT0008172
          View More Rows
          Pig ssc-miR-499-3p MIMAT0017373
          Pongo pygmaeus ppy-miR-499-3p MIMAT0015921
          Pteropus alecto pal-miR-499a-3p MIMAT0039821
          Rabbit ocu-miR-499-3p MIMAT0048490
          Rhesus monkey mml-miR-499-3p MIMAT0006362

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-499a
          Stem-loop Accession # MI0003183
          Stem-loop Sequence
          GCCCUGUCCCCUGUGCCUUGGGCGGGCGGCUGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCUACGCUGCCUGGGCAGGGU
          Chromosome Location Chr. 20 - 34990376 - 34990497 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004772
          miRBase ID: hsa-miR-499a-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: Chr. 20 - 34990376 - 34990497 [+] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-499
          Stem-loop Accession # MI0003721
          Stem-loop Sequence
          GCUGUUAAGACUUGCAGUGAUGUUUAGCUCCUCUCCAUGUGAACAUCACAGCAAGUCUGUGCUGC
          Chromosome Location Chr. 3 - 151138862 - 151138926 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017227
          miRBase ID: rno-miR-499-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: Chr. 3 - 151138862 - 151138926 [+] on Build Rnor_6.0

          Dasypus novemcinctus

          Stem-loop ID dno-mir-499
          Stem-loop Accession # MI0039134
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAACUCCUCUGCAUGUGAACAUCACAGCAAGUCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047937
          miRBase ID: dno-miR-499-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-499
          Stem-loop Accession # MI0038798
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAAUUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047298
          miRBase ID: cpo-miR-499-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-499
          Stem-loop Accession # MI0012858
          Stem-loop Sequence
          UGUUCUGUCCCCUGCACCCUGGGCGGGCGGCUGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCCAUGCUGCCUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013109
          miRBase ID: eca-miR-499-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-499
          Stem-loop Accession # MI0008694
          Stem-loop Sequence
          CCCUGUCCCCUGUGCCUUGGGCGGGCAGCUGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCUACGCUGCCUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008172
          miRBase ID: ptr-miR-499
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-499
          Stem-loop Accession # MI0013096
          Stem-loop Sequence
          UGGGCGGGCGGCCGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017373
          miRBase ID: ssc-miR-499-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-499
          Stem-loop Accession # MI0014972
          Stem-loop Sequence
          GCCCUGUCCCCUGUGCCUUGGGCGGGCGGCCGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCUACGCUGCUUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015921
          miRBase ID: ppy-miR-499-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-499a
          Stem-loop Accession # MI0032346
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039821
          miRBase ID: pal-miR-499a-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-499
          Stem-loop Accession # MI0039427
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAAUUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048490
          miRBase ID: ocu-miR-499-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-499
          Stem-loop Accession # MI0007773
          Stem-loop Sequence
          GCCCUGUCCCCGUGUCUUGGGCGGGCAGCUGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCUACGCUGCCUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006362
          miRBase ID: mml-miR-499-3p
          Mature miRNA Sequence: AACAUCACAGCAAGUCUGUGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304133_pri
          TaqMan™ Pri-miRNA Assay : Rn03464686_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12531
          Ambion® Pre-miR™ miRNA Precursor : PM12531
          TaqMan™ MicroRNA Assay : 002427
          mirVana® miRNA inhibitor : MH12531
          TaqMan™ Advanced miRNA Assay : rno478948_mir
          TaqMan™ Advanced miRNA Assay : 478948_mir


          miRBase Alias:

          Human: hsa-miR-499-3p (v17)
          Rat: rno-miR-499* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline