Search
Search

| Assay Name: | hsa-miR-499a-3p |
|---|---|
| miRBase Accession Number: | MI0003183 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACAUCACAGCAAGUCUGUGCU |
| Species: | Human, Rat, Dasypus novemcinctus, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | AACAUCACAGCAAGUCUGUGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-499a-3p | MIMAT0004772 |
| Rat | rno-miR-499-3p | MIMAT0017227 |
| Dasypus novemcinctus | dno-miR-499-3p | MIMAT0047937 |
| Guinea pig | cpo-miR-499-3p | MIMAT0047298 |
| Horse | eca-miR-499-3p | MIMAT0013109 |
| Pan troglodytes | ptr-miR-499 | MIMAT0008172 |
| View More Rows | ||
| Pig | ssc-miR-499-3p | MIMAT0017373 |
| Pongo pygmaeus | ppy-miR-499-3p | MIMAT0015921 |
| Pteropus alecto | pal-miR-499a-3p | MIMAT0039821 |
| Rabbit | ocu-miR-499-3p | MIMAT0048490 |
| Rhesus monkey | mml-miR-499-3p | MIMAT0006362 |