Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12542
          Assay Name: mmu-miR-130b-5p
          miRBase Accession Number: MI0000408
          miRBase Version: v22.1
          Chromosome Location: Chr.16: 17124061 - 17124142 [-] on Build GRCm38
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Species: Mouse, Rat, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Oreochromis niloticus, Papio hamadryas, Pig, Pundamilia nyererei, Rabbit, Rhesus monkey, Salmo salar, Xenopus laevis
          Product Type: mirVana® miRNA mimic
          Assay ID MC12542

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000034, mir-130

          Mature miRNA Sequence:

          ACUCUUUCCCUGUUGCACUACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-130b-5p MIMAT0004583
          Rat rno-miR-130b-5p MIMAT0017122
          Chrysemys picta cpi-miR-130b-5p MIMAT0037743
          Cricetulus griseus cgr-miR-130b-5p MIMAT0023756
          Dasypus novemcinctus dno-miR-130b-5p MIMAT0047629
          Goat chi-miR-130b-5p MIMAT0035945
          View More Rows
          Guinea pig cpo-miR-130b-5p MIMAT0046982
          Oreochromis niloticus oni-miR-130b-5p MIMAT0042825
          Papio hamadryas pha-miR-130b MIMAT0049550
          Pig ssc-miR-130b-5p MIMAT0037366
          Pundamilia nyererei pny-miR-130b-5p MIMAT0043017
          Rabbit ocu-miR-130b-5p MIMAT0048219
          Rhesus monkey mml-miR-130b-5p MIMAT0026815
          Salmo salar ssa-miR-130a-5p MIMAT0032321
          Xenopus laevis xla-miR-130b-5p MIMAT0046443

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-130b
          Stem-loop Accession # MI0000408
          Stem-loop Sequence
          GGCUUGUUGGACACUCUUUCCCUGUUGCACUACUGUGGGCCUCUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCUGUCGGGCC
          Chromosome Location Chr. 16 - 17124061 - 17124142 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004583
          miRBase ID: mmu-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: Chr. 16 - 17124061 - 17124142 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-130b
          Stem-loop Accession # MI0000904
          Stem-loop Sequence
          GGCUUGCUGGACACUCUUUCCCUGUUGCACUACUGUGGGCCUCUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCCGUCAGGCC
          Chromosome Location Chr. 11 - 88129773 - 88129854 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017122
          miRBase ID: rno-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: Chr. 11 - 88129773 - 88129854 [+] on Build Rnor_6.0

          Chrysemys picta

          Stem-loop ID cpi-mir-130b
          Stem-loop Accession # MI0029468
          Stem-loop Sequence
          CAGGCAGGCUGCCACUCUUUCCCUGUUGCACUACUGUUAAACUUGUAGCUAGCAGUGCAAUAAUGAAAGGGCGUCGGUCUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037743
          miRBase ID: cpi-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-130b
          Stem-loop Accession # MI0020395
          Stem-loop Sequence
          CUUGCUGGACACUCUUUCCCUGUUGCACUACUGUGGGCCACUGGGGAGCAGUGCAAUGAUGAAAGGGCAUCUGUCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023756
          miRBase ID: cgr-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-130b
          Stem-loop Accession # MI0038975
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGCCGCUGGGAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047629
          miRBase ID: dno-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-130b
          Stem-loop Accession # MI0030617
          Stem-loop Sequence
          GGCCUGCCUGACACUCUUUCCCUGUUGCACUACUGUGCGCCCCUGGCAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCGGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035945
          miRBase ID: chi-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-130b
          Stem-loop Accession # MI0038634
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGCUACUGGGAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046982
          miRBase ID: cpo-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-130b
          Stem-loop Accession # MI0034765
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGAGAUGCAGCAAGCAGUGCAAUAAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042825
          miRBase ID: oni-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-130b
          Stem-loop Accession # MI0040318
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGCCACUGGGAAGCAGUGCAAUGAUGAAAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049550
          miRBase ID: pha-miR-130b
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-130b
          Stem-loop Accession # MI0013136
          Stem-loop Sequence
          GCCUGCCUGACACUCUUUCCCUGUUGCACUACUGUGGGCCACUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCAGUCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037366
          miRBase ID: ssc-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-130b
          Stem-loop Accession # MI0035021
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGAGAUGCAGCAAGCAGUGCAAUAAUGAAAGGGCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0043017
          miRBase ID: pny-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-130b
          Stem-loop Accession # MI0039286
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGACCACUGGGAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048219
          miRBase ID: ocu-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-130b
          Stem-loop Accession # MI0007619
          Stem-loop Sequence
          GGCCUGCCCGACACUCUUUCCCUGUUGCACUACUGUGGGCCACUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCAGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026815
          miRBase ID: mml-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-130a-1
          Stem-loop Accession # MI0026468
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGAGCUGUAGCUAGCAGUGCAACAAUGAAAGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032321
          miRBase ID: ssa-miR-130a-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-130a-2
          Stem-loop Accession # MI0026469
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGAGCUGUAGCUAGCAGUGCAAUAAUGAAAGGGCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032321
          miRBase ID: ssa-miR-130a-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-130b
          Stem-loop Accession # MI0038245
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGCCAGUGAAUAAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046443
          miRBase ID: xla-miR-130b-5p
          Mature miRNA Sequence: ACUCUUUCCCUGUUGCACUACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465054_pri
          TaqMan™ Pri-miRNA Assay : Mm03306685_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12542
          Ambion® Pre-miR™ miRNA Precursor : PM12542
          TaqMan™ MicroRNA Assay : 002460
          mirVana® miRNA inhibitor : MH12542
          TaqMan™ Advanced miRNA Assay : rno480918_mir
          TaqMan™ Advanced miRNA Assay : mmu480918_mir


          miRBase Alias:

          Mouse: mmu-miR-130b* (v17)
          Rat: rno-miR-130b* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline