Search
Search

| Assay Name: | mmu-miR-130b-5p |
|---|---|
| miRBase Accession Number: | MI0000408 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUCUUUCCCUGUUGCACUACU |
| Species: | Mouse, Rat, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Oreochromis niloticus, Papio hamadryas, Pig, Pundamilia nyererei, Rabbit, Rhesus monkey, Salmo salar, Xenopus laevis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | ACUCUUUCCCUGUUGCACUACU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-130b-5p | MIMAT0004583 |
| Rat | rno-miR-130b-5p | MIMAT0017122 |
| Chrysemys picta | cpi-miR-130b-5p | MIMAT0037743 |
| Cricetulus griseus | cgr-miR-130b-5p | MIMAT0023756 |
| Dasypus novemcinctus | dno-miR-130b-5p | MIMAT0047629 |
| Goat | chi-miR-130b-5p | MIMAT0035945 |
| View More Rows | ||
| Guinea pig | cpo-miR-130b-5p | MIMAT0046982 |
| Oreochromis niloticus | oni-miR-130b-5p | MIMAT0042825 |
| Papio hamadryas | pha-miR-130b | MIMAT0049550 |
| Pig | ssc-miR-130b-5p | MIMAT0037366 |
| Pundamilia nyererei | pny-miR-130b-5p | MIMAT0043017 |
| Rabbit | ocu-miR-130b-5p | MIMAT0048219 |
| Rhesus monkey | mml-miR-130b-5p | MIMAT0026815 |
| Salmo salar | ssa-miR-130a-5p | MIMAT0032321 |
| Xenopus laevis | xla-miR-130b-5p | MIMAT0046443 |