Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12582
          Assay Name: hsa-miR-125b-1-3p
          miRBase Accession Number: MI0000446
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 122099757 - 122099844 [-] on Build GRCh38
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Columba livia, Dasypus novemcinctus, Eptesicus fuscus, Guinea pig, Ophiophagus hannah, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12582

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          ACGGGUUAGGCUCUUGGGAGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-125b-1-3p MIMAT0004592
          Mouse mmu-miR-125b-1-3p MIMAT0004669
          Rat rno-miR-125b-1-3p MIMAT0004730
          Alligator mississippiensis ami-miR-125b-3p MIMAT0038129
          Columba livia cli-miR-125-3p MIMAT0038497
          Dasypus novemcinctus dno-miR-125b-3p MIMAT0047615
          View More Rows
          Eptesicus fuscus efu-miR-125a MIMAT0035053
          Guinea pig cpo-miR-125b-3p MIMAT0046968
          Ophiophagus hannah oha-miR-125b-3p MIMAT0036675
          Rabbit ocu-miR-125b-3p MIMAT0048206
          Rhesus monkey mml-miR-125b-1-3p MIMAT0026564

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-125b-1
          Stem-loop Accession # MI0000446
          Stem-loop Sequence
          UGCGCUCCUCUCAGUCCCUGAGACCCUAACUUGUGAUGUUUACCGUUUAAAUCCACGGGUUAGGCUCUUGGGAGCUGCGAGUCGUGCU
          Chromosome Location Chr. 11 - 122099757 - 122099844 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004592
          miRBase ID: hsa-miR-125b-1-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: Chr. 11 - 122099757 - 122099844 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-125b-1
          Stem-loop Accession # MI0000725
          Stem-loop Sequence
          UGCGCUCCCCUCAGUCCCUGAGACCCUAACUUGUGAUGUUUACCGUUUAAAUCCACGGGUUAGGCUCUUGGGAGCUG
          Chromosome Location Chr. 9 - 41581926 - 41582002 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004669
          miRBase ID: mmu-miR-125b-1-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: Chr. 9 - 41581926 - 41582002 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-125b-1
          Stem-loop Accession # MI0000896
          Stem-loop Sequence
          UGCGCUCCCCUCAGUCCCUGAGACCCUAACUUGUGAUGUUUACCGUUUAAAUCCACGGGUUAGGCUCUUGGGAGCUGCGAGUCGUGC
          Chromosome Location Chr. 8 - 45798260 - 45798346 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004730
          miRBase ID: rno-miR-125b-1-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: Chr. 8 - 45798260 - 45798346 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-125b-1
          Stem-loop Accession # MI0029722
          Stem-loop Sequence
          UUUGUGCGCCCCUCUCAAUCCCUGAGACCCUAACUUGUGAUGUUUAGCUUUUAAAUCCACGGGUUAGGCUCUUGGGAGCUGUGAGUUGUGCUUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038129
          miRBase ID: ami-miR-125b-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-125-1
          Stem-loop Accession # MI0029956
          Stem-loop Sequence
          GUUGCGCCCCUCUCAAUCCCUGAGACCCUAACUUGUGAUGUUUAGCUUUUAAAUCCACGGGUUAGGCUCUUGGGAGCUGUGAGUUGUGCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038497
          miRBase ID: cli-miR-125-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-125b-1
          Stem-loop Accession # MI0038966
          Stem-loop Sequence
          UCCCUGAGACCCUAACUUGUGAUGUUUACCGUUUAAAUCCACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047615
          miRBase ID: dno-miR-125b-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-125a
          Stem-loop Accession # MI0028740
          Stem-loop Sequence
          GCCCCUCUCGAUCCCUGAGACCCUAACUUGUGAUGUUUUCCGUUUAAAUCCACGGGUUAGGCUCUUGGGAGCUGCGAGCCGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035053
          miRBase ID: efu-miR-125a
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-125b-1
          Stem-loop Accession # MI0038625
          Stem-loop Sequence
          UCCCUGAGACCCUAACUUGUGAUGUUUACCGUUUAAAUCCACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046968
          miRBase ID: cpo-miR-125b-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-125b
          Stem-loop Accession # MI0031328
          Stem-loop Sequence
          GGACUUGCGCCCCUCUCUGUCCCUGAGACCCUAACUUGUGACGUUUUGGUUUCAAGUCCACGGGUUAGGCUCUUGGGAGCUGUGAGUUGUGCCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036675
          miRBase ID: oha-miR-125b-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-125b-1
          Stem-loop Accession # MI0039278
          Stem-loop Sequence
          UCCCUGAGACCCUAACUUGUGAUGUUUACCGUUUAAAUCCACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048206
          miRBase ID: ocu-miR-125b-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-125b-1
          Stem-loop Accession # MI0002514
          Stem-loop Sequence
          UGCGCUCCUCUCAGUCCCUGAGACCCUAACUUGUGAUGUUUACCGUUUAAAUCCACGGGUUAGGCUCUUGGGAGCUGCGAGUCGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026564
          miRBase ID: mml-miR-125b-1-3p
          Mature miRNA Sequence: ACGGGUUAGGCUCUUGGGAGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465285_pri
          TaqMan™ Pri-miRNA Assay : Hs03303095_pri
          TaqMan™ Pri-miRNA Assay : Mm03307273_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12582
          Ambion® Pre-miR™ miRNA Precursor : PM12582
          TaqMan™ MicroRNA Assay : 002378
          mirVana® miRNA inhibitor : MH12582
          TaqMan™ Advanced miRNA Assay : mmu481296_mir
          TaqMan™ Advanced miRNA Assay : 478665_mir
          TaqMan™ Advanced miRNA Assay : rno481296_mir


          miRBase Alias:

          Human: hsa-miR-125b-1* (v17)
          Rat: rno-miR-125b-3p (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0