Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13132
          Assay Name: hsa-miR-320b
          miRBase Accession Number: MI0003776
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 116671746 - 116671817 [+] on Build GRCh38
          Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCAA
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC13132

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000163, mir-320

          Mature miRNA Sequence:

          AAAAGCUGGGUUGAGAGGGCAA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-320b MIMAT0005792
          Pan troglodytes ptr-miR-320b MIMAT0008094
          Pongo pygmaeus ppy-miR-320b MIMAT0015822
          Rhesus monkey mml-miR-320b MIMAT0024312

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-320b-1
          Stem-loop Accession # MI0003776
          Stem-loop Sequence
          AUAAAUUAAUCCCUCUCUUUCUAGUUCUUCCUAGAGUGAGGAAAAGCUGGGUUGAGAGGGCAAACAAAUUAA
          Chromosome Location Chr. 1 - 116671746 - 116671817 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005792
          miRBase ID: hsa-miR-320b
          Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCAA
          Chromosome Location: Chr. 1 - 116671746 - 116671817 [+] on Build GRCh38

          Stem-loop ID hsa-mir-320b-2
          Stem-loop Accession # MI0003839
          Stem-loop Sequence
          GUCUCUUAGGCUUUCUCUUCCCAGAUUUCCCAAAGUUGGGAAAAGCUGGGUUGAGAGGGCAAAAGGAAAAA
          Chromosome Location Chr. 1 - 224257040 - 224257110 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005792
          miRBase ID: hsa-miR-320b
          Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCAA
          Chromosome Location: Chr. 1 - 116671746 - 116671817 [+] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-320b-1
          Stem-loop Accession # MI0008608
          Stem-loop Sequence
          AAUUAAUCCCUCUCUUUCUAGUUCUUCCUAGAGUGAGGAAAAGCUGGGUUGAGAGGGCAAACAAAUUAACUAAUUAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008094
          miRBase ID: ptr-miR-320b
          Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCAA
          Chromosome Location: NA

          Stem-loop ID ptr-mir-320b-2
          Stem-loop Accession # MI0008609
          Stem-loop Sequence
          UGUUAUUUUUUGUCUUCUACCUAAGAAUUCUGUCUCUUAGGCUUUCUCUUCCCAGAUUUCCCAAAGUUGGGAAAAGCUGGGUUGAGAGGGCAAAAGGAAAAAAAAAGAAUUCUGUCUCUGACAUAAUUAGAUAGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008094
          miRBase ID: ptr-miR-320b
          Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCAA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-320b-1
          Stem-loop Accession # MI0014885
          Stem-loop Sequence
          AAUUAAUCCCUCUCUUUCUAGUUCUUCCUAGAGUAAGGAAAAGCUGGGUUGAGAGGGCAAACAAAUUAACUAAUUAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015822
          miRBase ID: ppy-miR-320b
          Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCAA
          Chromosome Location: NA

          Stem-loop ID ppy-mir-320b-2
          Stem-loop Accession # MI0014886
          Stem-loop Sequence
          UGUUAUUUUUUGUCUUCUACCUAAGAAUUCUGUCUCUUAGGCUUUCUCUUCUCAGUUUUCCCAAAGUUGAGAAAAGCUGGGUUGAGAGGGCAAAAGGAAAAAAAAAGAAUUCUGUCUCUGACACAAUUAGAUAGGGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015822
          miRBase ID: ppy-miR-320b
          Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCAA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-320b
          Stem-loop Accession # MI0020860
          Stem-loop Sequence
          GUUAUUUUUAGUCUUCUACCUAAGAAUUCUGUCUCUUAGGCUUUCUCUUCCCAGUUUUCCCAAAGUUGGGAAAAGCUGGGUUGAGAGGGCAAAAGGAAAAAGAAAAAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024312
          miRBase ID: mml-miR-320b
          Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCAA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304949_pri
          TaqMan™ Pri-miRNA Assay : Hs03305009_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13132
          Ambion® Pre-miR™ miRNA Precursor : PM13132
          TaqMan™ MicroRNA Assay : 002844
          mirVana® miRNA inhibitor : MH13132
          TaqMan™ Advanced miRNA Assay : 478588_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline