Search
Search

| Assay Name: | hsa-miR-320B |
|---|---|
| miRBase Accession Number: | MI0003776 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCAA |
| Species: | Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-320b | MIMAT0005792 |
| Pan troglodytes | ptr-miR-320b | MIMAT0008094 |
| Pongo pygmaeus | ppy-miR-320b | MIMAT0015822 |
| Rhesus monkey | mml-miR-320b | MIMAT0024312 |