Search
Search

| Assay Name: | oan-miR-194-3p |
|---|---|
| miRBase Accession Number: | MI0006795 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCAGUGGAGCUGCUGUUAUCUU |
| Species: | Ornithorhynchus anatinus |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000055, mir-194 |
Mature miRNA Sequence: | CCAGUGGAGCUGCUGUUAUCUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-194-3p | MIMAT0007018 |