Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC14836
          Assay Name: mmu-miR-133b-5p
          miRBase Accession Number: MI0000821
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 20682769 - 20682887 [+] on Build GRCm38
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Species: Mouse, Dasypus novemcinctus, Gadus morhua, Guinea pig, Monodelphis domestica, Ornithorhynchus anatinus, Paralichthys olivaceus, Rabbit, Salmo salar
          Product Type: mirVana® miRNA mimic
          Assay ID MC14836

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000029, mir-133

          Mature miRNA Sequence:

          GCUGGUCAAACGGAACCAAGUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-133b-5p MIMAT0017083
          Dasypus novemcinctus dno-miR-133b-5p MIMAT0047635
          Gadus morhua gmo-miR-133b-5p MIMAT0044200
          Guinea pig cpo-miR-133b-5p MIMAT0046988
          Monodelphis domestica mdo-miR-133b-5p MIMAT0050179
          Ornithorhynchus anatinus oan-miR-133b-5p MIMAT0006996
          View More Rows
          Paralichthys olivaceus pol-miR-133-5p MIMAT0025432
          Rabbit ocu-miR-133b-5p MIMAT0048223
          Salmo salar ssa-miR-133b-5p MIMAT0032339

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-133b
          Stem-loop Accession # MI0000821
          Stem-loop Sequence
          CCUCCAAAGGGAGUGGCCCCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAAAGCUCAAUAUUUGGAGA
          Chromosome Location Chr. 1 - 20682769 - 20682887 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017083
          miRBase ID: mmu-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: Chr. 1 - 20682769 - 20682887 [+] on Build GRCm38

          Dasypus novemcinctus

          Stem-loop ID dno-mir-133b
          Stem-loop Accession # MI0038979
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCCGUCUUCUUGAGAGGUUUGGUCCCCUUCAACCAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047635
          miRBase ID: dno-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-133b
          Stem-loop Accession # MI0036117
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCAGGUGUUUCUGUGAGGUUUGGUCCCUUUCAACCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044200
          miRBase ID: gmo-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-133b
          Stem-loop Accession # MI0038638
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046988
          miRBase ID: cpo-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-133b
          Stem-loop Accession # MI0040828
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCCGUCUGACUGAGAGGUUUGGUCCCCUUCAACCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0050179
          miRBase ID: mdo-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-133b
          Stem-loop Accession # MI0006780
          Stem-loop Sequence
          CGGCUGAACCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUGCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCGGGGCCGACAUCGUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006996
          miRBase ID: oan-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-133
          Stem-loop Accession # MI0022199
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCAGGUGUUUCUGUGAGGUUUGGUCCCCUUCAACCAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025432
          miRBase ID: pol-miR-133-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-133b
          Stem-loop Accession # MI0039288
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048223
          miRBase ID: ocu-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-133b-1
          Stem-loop Accession # MI0026481
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCAGGUGUUUUAGUGAGGUUUGGUCCCCUUCAACCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032339
          miRBase ID: ssa-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-133b-2
          Stem-loop Accession # MI0026482
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCAGGUGUUUCAGUGAGGUUUGGUCCCCUUCAACCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032339
          miRBase ID: ssa-miR-133b-5p
          Mature miRNA Sequence: GCUGGUCAAACGGAACCAAGUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307410_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM14836
          Ambion® Pre-miR™ miRNA Precursor : PM14836
          TaqMan™ MicroRNA Assay : 007577_mat
          mirVana® miRNA inhibitor : MH14836
          TaqMan™ Advanced miRNA Assay : mmu481504_mir


          miRBase Alias:

          Mouse: mmu-miR-133b* (v17)
          Ochrobactrum anthropi: oan-miR-133b* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline