Search
Search

| Assay Name: | mmu-miR-133b-5p |
|---|---|
| miRBase Accession Number: | MI0000821 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCUGGUCAAACGGAACCAAGUC |
| Species: | Mouse, Dasypus novemcinctus, Gadus morhua, Guinea pig, Monodelphis domestica, Ornithorhynchus anatinus, Paralichthys olivaceus, Rabbit, Salmo salar |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000029, mir-133 |
Mature miRNA Sequence: | GCUGGUCAAACGGAACCAAGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-133b-5p | MIMAT0017083 |
| Dasypus novemcinctus | dno-miR-133b-5p | MIMAT0047635 |
| Gadus morhua | gmo-miR-133b-5p | MIMAT0044200 |
| Guinea pig | cpo-miR-133b-5p | MIMAT0046988 |
| Monodelphis domestica | mdo-miR-133b-5p | MIMAT0050179 |
| Ornithorhynchus anatinus | oan-miR-133b-5p | MIMAT0006996 |
| View More Rows | ||
| Paralichthys olivaceus | pol-miR-133-5p | MIMAT0025432 |
| Rabbit | ocu-miR-133b-5p | MIMAT0048223 |
| Salmo salar | ssa-miR-133b-5p | MIMAT0032339 |