Search
Search

| Assay Name: | cfa-miR-615 |
|---|---|
| miRBase Accession Number: | MI0010419 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGGGGUCCCCGGAGCUCGG |
| Species: | Dog |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000342, mir-615 |
Mature miRNA Sequence: | GGGGGUCCCCGGAGCUCGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-615 | MIMAT0009920 |