Search
Search

| Assay Name: | rno-miR-153-5p |
|---|---|
| miRBase Accession Number: | MI0000922 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUCAUUUUUGUGAUGUUGCAGCU |
| Species: | Rat, Columba livia, Dasypus novemcinctus, Guinea pig, Ophiophagus hannah, Pteropus alecto, Python bivittatus, Rabbit, Taeniopygia guttata, Zebrafish |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | GUCAUUUUUGUGAUGUUGCAGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-153-5p | MIMAT0017135 |
| Columba livia | cli-miR-153b-5p | MIMAT0038547 |
| Dasypus novemcinctus | dno-miR-153b-5p | MIMAT0047681 |
| Guinea pig | cpo-miR-153-5p | MIMAT0047034 |
| Ophiophagus hannah | oha-miR-153-1-5p | MIMAT0036737 |
| Pteropus alecto | pal-miR-153-5p | MIMAT0040065 |
| View More Rows | ||
| Python bivittatus | pbv-miR-153-5p | MIMAT0038970 |
| Rabbit | ocu-miR-153-5p | MIMAT0048269 |
| Taeniopygia guttata | tgu-miR-153-1-5p | MIMAT0026997 |
| Zebrafish | dre-miR-153a-5p | MIMAT0031978 |