Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno480950_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-153-5p
          Stem-loop Accession Number: MI0000922
          miRBase Version: v22.1
          Chromosome Location: Chr.6: 145105064 - 145105150 [+] on Build Rnor_6.0
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Species: Rat, Columba livia, Dasypus novemcinctus, Guinea pig, Ophiophagus hannah, Pteropus alecto, Python bivittatus, Rabbit, Taeniopygia guttata, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno480950_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000050, mir-153

          Mature miRNA Sequence:

          GUCAUUUUUGUGAUGUUGCAGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-153-5p MIMAT0017135
          Columba livia cli-miR-153b-5p MIMAT0038547
          Dasypus novemcinctus dno-miR-153b-5p MIMAT0047681
          Guinea pig cpo-miR-153-5p MIMAT0047034
          Ophiophagus hannah oha-miR-153-1-5p MIMAT0036737
          Pteropus alecto pal-miR-153-5p MIMAT0040065
          View More Rows
          Python bivittatus pbv-miR-153-5p MIMAT0038970
          Rabbit ocu-miR-153-5p MIMAT0048269
          Taeniopygia guttata tgu-miR-153-1-5p MIMAT0026997
          Zebrafish dre-miR-153a-5p MIMAT0031978

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-153
          Stem-loop Accession # MI0000922
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location Chr. 6 - 145105064 - 145105150 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017135
          miRBase ID: rno-miR-153-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: Chr. 6 - 145105064 - 145105150 [+] on Build Rnor_6.0

          Columba livia

          Stem-loop ID cli-mir-153b
          Stem-loop Accession # MI0029987
          Stem-loop Sequence
          UUAGCGGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038547
          miRBase ID: cli-miR-153b-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-153b
          Stem-loop Accession # MI0039003
          Stem-loop Sequence
          GUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047681
          miRBase ID: dno-miR-153b-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-153
          Stem-loop Accession # MI0038662
          Stem-loop Sequence
          GUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047034
          miRBase ID: cpo-miR-153-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-153-1
          Stem-loop Accession # MI0031364
          Stem-loop Sequence
          UGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUAAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036737
          miRBase ID: oha-miR-153-1-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-153-1
          Stem-loop Accession # MI0032475
          Stem-loop Sequence
          GUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040065
          miRBase ID: pal-miR-153-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-153-1
          Stem-loop Accession # MI0030250
          Stem-loop Sequence
          UAGCAGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUAAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038970
          miRBase ID: pbv-miR-153-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-153
          Stem-loop Accession # MI0039312
          Stem-loop Sequence
          GUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048269
          miRBase ID: ocu-miR-153-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-153-1
          Stem-loop Accession # MI0013768
          Stem-loop Sequence
          UGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026997
          miRBase ID: tgu-miR-153-1-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-153a
          Stem-loop Accession # MI0002021
          Stem-loop Sequence
          GGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUUAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031978
          miRBase ID: dre-miR-153a-5p
          Mature miRNA Sequence: GUCAUUUUUGUGAUGUUGCAGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465317_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19787
          Ambion® Pre-miR™ miRNA Precursor : PM19787
          TaqMan™ MicroRNA Assay : 461783_mat
          mirVana® miRNA inhibitor : MH19787
          mirVana® miRNA mimic : MC19787

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline