Search
Search

| Assay Name: | rno-miR-134-3p |
|---|---|
| miRBase Accession Number: | MI0000907 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGUGGGCCACCUAGUCACCAA |
| Species: | Rat, Guinea pig |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000112, mir-134 |
Mature miRNA Sequence: | CUGUGGGCCACCUAGUCACCAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-134-3p | MIMAT0017125 |
| Guinea pig | cpo-miR-134-3p | MIMAT0046991 |