Search
Search

| Assay Name: | chi-miR-361-3p |
|---|---|
| miRBase Accession Number: | MI0030760 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCCCCAGGUGUGAUUCUGAUU |
| Species: | Goat |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000172, mir-361 |
Mature miRNA Sequence: | UCCCCCAGGUGUGAUUCUGAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Goat | chi-miR-361-3p | MIMAT0036172 |