Search
Search

| Assay Name: | hsa-miR-598-3p |
|---|---|
| miRBase Accession Number: | MI0003610 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACGUCAUCGUUGUCAUCGUCA |
| Species: | Human, Gorilla gorilla, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000393, mir-598 |
Mature miRNA Sequence: | UACGUCAUCGUUGUCAUCGUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-598-3p | MIMAT0003266 |
| Gorilla gorilla | ggo-miR-598 | MIMAT0024089 |
| Horse | eca-miR-598 | MIMAT0012922 |
| Pan troglodytes | ptr-miR-598 | MIMAT0008267 |
| Pongo pygmaeus | ppy-miR-598 | MIMAT0016040 |
| Rhesus monkey | mml-miR-598-3p | MIMAT0006455 |