Search
Search

| Assay Name: | ath-miR319c |
|---|---|
| miRBase Accession Number: | MI0001086 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGGACUGAAGGGAGCUCCUU |
| Species: | Arabidopsis, Arabidopsis lyrata, Manihot esculenta, Phaseolus vulgaris, Ricinus communis, Solanum lycopersicum |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000010, MIR159 |
Mature miRNA Sequence: | UUGGACUGAAGGGAGCUCCUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR319c | MIMAT0001016 |
| Arabidopsis lyrata | aly-miR319c-3p | MIMAT0017434 |
| Arabidopsis lyrata | aly-miR319d-3p | MIMAT0017664 |
| Manihot esculenta | mes-miR319g | MIMAT0029259 |
| Phaseolus vulgaris | pvu-miR319c | MIMAT0011176 |
| Ricinus communis | rco-miR319d | MIMAT0014183 |
| View More Rows | ||
| Solanum lycopersicum | sly-miR319c-3p | MIMAT0035432 |