Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12000
          Assay Name: ath-miR319c
          miRBase Accession Number: MI0001086
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 17029701 - 17029899 [+] on Build TAIR10.1
          Mature miRNA Sequence: UUGGACUGAAGGGAGCUCCUU
          Species: Arabidopsis, Arabidopsis lyrata, Manihot esculenta, Phaseolus vulgaris, Ricinus communis, Solanum lycopersicum
          Product Type: mirVana® miRNA mimic
          Assay ID MC12000

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000010, MIR159

          Mature miRNA Sequence:

          UUGGACUGAAGGGAGCUCCUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Arabidopsis ath-miR319c MIMAT0001016
          Arabidopsis lyrata aly-miR319c-3p MIMAT0017434
          Arabidopsis lyrata aly-miR319d-3p MIMAT0017664
          Manihot esculenta mes-miR319g MIMAT0029259
          Phaseolus vulgaris pvu-miR319c MIMAT0011176
          Ricinus communis rco-miR319d MIMAT0014183
          View More Rows
          Solanum lycopersicum sly-miR319c-3p MIMAT0035432

          Stem-loop Details



          Arabidopsis

          Stem-loop ID ath-MIR319c
          Stem-loop Accession # MI0001086
          Stem-loop Sequence
          UAGAUAUAGAAGGAGAUUCUUUCAGUCCAGUCAUGGAUAGAAAAAGAAGAGGGUAGAAAUAUCUGCCGACUCAUCCAUCCAAACACUCGUGGUAGAGAAACGAUAAAUUUAAACCGCAGUGACUGUGUGAAUGAUGCGGGAGAUAUUUUUGAUCCUUCUUUAUCUGUGUUUGGACUGAAGGGAGCUCCUUCUUUUUCUA
          Chromosome Location Chr. 2 - 17029701 - 17029899 [+] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0001016
          miRBase ID: ath-miR319c
          Mature miRNA Sequence: UUGGACUGAAGGGAGCUCCUU
          Chromosome Location: Chr. 2 - 17029701 - 17029899 [+] on Build TAIR10.1

          Arabidopsis lyrata

          Stem-loop ID aly-MIR319c
          Stem-loop Accession # MI0014520
          Stem-loop Sequence
          AUAGAUAAAUAUAUGAAUCGAGAUUAAAAUAGAUAGAUAUAGAAGGAGAUUCUUUCAGUCCAGUCAUGGAUAGAAAAAGAAGAGGGUAGAAAUAUCUGCCGACUCAUCCAUCCAAACACUCGUGGUAGAGAAACGAUCAAUUUUAAACCGCAGUGACUGUAUGAAUGAUGCGGGAGAUAUUUUUGAUCCUUCUUUAUCUGUGUUUGGACUGAAGGGAGCUCCUUCUUUUUCUACAAUUUUUUCAUUUUUUUCUCAUUACCUGUUUAUAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017434
          miRBase ID: aly-miR319c-3p
          Mature miRNA Sequence: UUGGACUGAAGGGAGCUCCUU
          Chromosome Location: NA

          Stem-loop ID aly-MIR319d
          Stem-loop Accession # MI0014633
          Stem-loop Sequence
          AUAGAUAGAUAUAUGAAUCGAGAUUAAAAUAGAUAGAUAUAGAAGGAGAUUCUUUCAGUCCAGUCAUGGAUAGAAAAAGAAGAGGGUAGAAAUAUCUGCCGACUCAUCCAUCCAAACACUCGUGGUAGAGAAACGAUCAAUUUUAAACCGCAGUGACUGUAUGAAUGAUGCGGGAGAUAUUUUUGAUCCUUCUUUAUCUGUGUUUGGACUGAAGGGAGCUCCUUCUUUUUCUACAAUUUUUUCAUUUUUUUCUCAUCAUCUGUUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017664
          miRBase ID: aly-miR319d-3p
          Mature miRNA Sequence: UUGGACUGAAGGGAGCUCCUU
          Chromosome Location: NA

          Manihot esculenta

          Stem-loop ID mes-MIR319g
          Stem-loop Accession # MI0024301
          Stem-loop Sequence
          GAAUGUGGAAAUGGUGUGAGAGAGCUUCCUUCAGCCCACUCAUGGAUGGGGUAAAAGGGGCGGAAAUAUCUGCCGACUCAUUCAUUCAAGCACUAGUAGACAGAGAUACUGGUAAUACUGUGUUUAGCUACUGUGAUUGCGUGAAUGAUGCGGGAGAUAAUUUUGCAUCCCUCUUUUCUGUGCUUGGACUGAAGGGAGCUCCUUCCUUCUCUAUUACCUACAUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029259
          miRBase ID: mes-miR319g
          Mature miRNA Sequence: UUGGACUGAAGGGAGCUCCUU
          Chromosome Location: NA

          Phaseolus vulgaris

          Stem-loop ID pvu-MIR319c
          Stem-loop Accession # MI0010705
          Stem-loop Sequence
          GAGAGUGAAGGAGCUUCCCUCAGCCCAUUCAUGGAUACAAAAGAUUGGGUUGCAAAAUUAACCACUAACUCAUUCACACAAUAGUAUUCAGUUAGGGUGAUGCUCUUUUUGCUAUAAUAUGAUUGUGUGAAUGAAGUGAUGCAUACAUAUUGUGUGACCCUCUUUCUCUGAGCUUGGACUGAAGGGAGCUCCUUCUUUCUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011176
          miRBase ID: pvu-miR319c
          Mature miRNA Sequence: UUGGACUGAAGGGAGCUCCUU
          Chromosome Location: NA

          Ricinus communis

          Stem-loop ID rco-MIR319d
          Stem-loop Accession # MI0013409
          Stem-loop Sequence
          CCAAAGGAGCUCCUUUCAGUCCAAUAUGGAGGGCUUAGAAGCGGCAAAGAGCUGCCAUCUCAUGCAUUAAGGCAAUACUUGACAAGUUUUUCACAGUGAAAAUCUCAAGUUUGGCCUGGUGCAUGGUGUGGGAGCAACUCCUUCCCUCCUUCCCCCACCCAUUGGACUGAAGGGAGCUCCUUCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014183
          miRBase ID: rco-miR319d
          Mature miRNA Sequence: UUGGACUGAAGGGAGCUCCUU
          Chromosome Location: NA

          Solanum lycopersicum

          Stem-loop ID sly-MIR319c
          Stem-loop Accession # MI0029099
          Stem-loop Sequence
          GGAAUGGUCCAAGAGAGCUUCCUUCAGCCCACUCACGGAGGAGAAAUGGGGUUGAAUUAGCUGCCGACUCAUUCAUCCAACCACUCAGUAGAAAGGAUAGGUUUUGUGCUACUGUGAUUGAGUGAAUGAUGCGGGAGAUAGUUUUCUAUCCCUCUCUUUCCUUGCUUGGACUGAAGGGAGCUCCUUUUCCCCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035432
          miRBase ID: sly-miR319c-3p
          Mature miRNA Sequence: UUGGACUGAAGGGAGCUCCUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 004410_mat
          mirVana® miRNA inhibitor : MH12000


          miRBase Alias:

          Arabidopsis lyrata: aly-miR319d (v18)
          Arabidopsis lyrata: aly-miR319c (v18)
          Microbacterium esteraromaticum: mes-miR319f (v21)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline