Search
Search

| Assay Name: | hsa-miR-105-3p |
|---|---|
| miRBase Accession Number: | MI0000111 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACGGAUGUUUGAGCAUGUGCUA |
| Species: | Human, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000074, mir-105 |
Mature miRNA Sequence: | ACGGAUGUUUGAGCAUGUGCUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-105-3p | MIMAT0004516 |
| Rhesus monkey | mml-miR-105-3p | MIMAT0026583 |