Search
Search

| Assay Name: | hsa-miR-548am-5p |
|---|---|
| miRBase Accession Number: | MI0016904 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAAGUAAUUGCGGUUUUUGCC |
| Species: | Human, Pan troglodytes |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUAAUUGCGGUUUUUGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548am-5p | MIMAT0022740 |
| Human | hsa-miR-548c-5p | MIMAT0004806 |
| Human | hsa-miR-548o-5p | MIMAT0022738 |
| Pan troglodytes | ptr-miR-548o | MIMAT0024054 |