Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04257665_pri
          Assay Name:
          hsa-mir-548o-2
          Stem-loop ID:
          hsa-mir-548o-2
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.20: 38516563 - 38516632 [+] on Build GRCh38
          Stem-loop Location:
          Chr.20: 38516563 - 38516632 [+] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-548o-5p hsa-miR-548o-3p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs04257665_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          105

          Chromosome Location:

          Chr. 20 - 38516411 - 38516411 [+] on 38516411 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-548o-5p MIMAT0022738
          Human hsa-miR-548o-3p MIMAT0005919

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-548o-2
          Stem-loop Accession # MI0016746
          Stem-loop Sequence
          UGGUGCAAAAGUAAUUGCGGUUUUUGCCAUUAAAAGUAAUGCGGCCAAAACUGCAGUUACUUUUGCACCC
          Chromosome Location Chr. 20 - 38516563 - 38516632 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0022738
          miRBase ID: hsa-miR-548o-5p
          Mature miRNA Sequence: AAAAGUAAUUGCGGUUUUUGCC
          Chromosome Location: Chr. 20 - 38516563 - 38516632 [+] on Build GRCh38

          miRBase Accession #: MIMAT0005919
          miRBase ID: hsa-miR-548o-3p
          Mature miRNA Sequence: CCAAAACUGCAGUUACUUUUGC
          Chromosome Location: Chr. 20 - 38516563 - 38516632 [+] on Build GRCh38

          Gene Family ID MIPF0000317, mir-548
          Distance to Amplicon 106 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12894
          Ambion® Pre-miR™ miRNA Precursor : PM12894
          TaqMan™ MicroRNA Assay : 002429
          mirVana® miRNA inhibitor : MH12894
          mirVana® miRNA mimic : MC12894
          TaqMan™ Advanced miRNA Assay : 480872_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13332
          Ambion® Pre-miR™ miRNA Precursor : PM13332
          TaqMan™ MicroRNA Assay : 245470_mat
          mirVana® miRNA inhibitor : MH13332
          mirVana® miRNA mimic : MC13332

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline