Search
Search

| Assay Name: | hsa-miR-519b-3p |
|---|---|
| miRBase Accession Number: | MI0003151 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAGUGCAUCCUUUUAGAGGUU |
| Species: | Human, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCAUCCUUUUAGAGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-519b-3p | MIMAT0002837 |
| Gorilla gorilla | ggo-miR-519c-3p | MIMAT0036411 |
| Pan troglodytes | ptr-miR-519b | MIMAT0008193 |
| Pongo pygmaeus | ppy-miR-519b | MIMAT0015959 |