Search
Search

ZNF589| Gene Symbol: ZNF589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016089.2 | NP_057173.2 | 4 | 2691 | ||
| XM_017006575.1 | XP_016862064.1 | 4 | 2772 | |||
| XM_017006576.1 | XP_016862065.1 | 6 | 2921 | |||
| XM_017006577.1 | XP_016862066.1 | 4 | 3977 | |||
| XM_017006578.1 | XP_016862067.1 | 4 | 2752 | |||
| XM_017006579.1 | XP_016862068.1 | 6 | 2877 | |||
| GenBank mRNA | AB209387.1 | - | 4 | 2672 | ||
| AF114816.1 | - | 7 | 1710 | |||
| AF114817.1 | - | 6 | 2994 | |||
| AL832831.1 | - | 4 | 2678 | |||
| AY258146.1 | - | 4 | 2691 | |||
| BC005859.2 | - | 4 | 2650 | |||
| BC006247.2 | - | 4 | 2671 | |||
| BC008213.1 | - | 1 | 1259 | |||
| BC048798.1 | - | 4 | 2668 | |||