Search
Search

Nhp2l1
LOC100359574| Gene Symbol: Nhp2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212515.2 | NP_997680.1 | 2 | 191 | ||
| GenBank mRNA | BC058493.1 | - | 2 | 194 | 6887059 | |
| FQ220734.1 | - | 192 | ||||
| FQ233358.1 | - | 192 | ||||
| Gene Symbol: LOC100359574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760591.1 | XP_008758813.1 | 1 | 884 | ||