Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017508.1 | NP_001017508.1 | 6 | 1465 | ||
XM_008771406.1 | XP_008769628.1 | 6 | 8193 | |||
GenBank mRNA | BC079268.1 | - | 6 | 1465 | 7133664 |