Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013001.2 | NP_037133.1 | 5 | 344 | ||
XM_006234633.2 | XP_006234695.1 | 7 | 769 | |||
XM_006234634.2 | XP_006234696.1 | 7 | 2225 | |||
XM_006234635.2 | XP_006234697.1 | 7 | 684 | |||
XM_006234636.2 | XP_006234698.1 | 7 | 1675 | |||
XM_008762060.1 | XP_008760282.1 | 6 | 868 | |||
GenBank mRNA | AJ627631.1 | - | 3 | 226 | ||
AY540905.1 | - | 4 | 241 | |||
AY540906.1 | - | 4 | 241 | |||
BC128741.1 | - | 5 | 344 | 8363963 | ||
HQ121510.1 | - | 3 | 227 | |||
U69644.1 | - | 5 | 564 |