Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282992.1 | NP_001269921.1 | 2 | 387 | ||
NM_008787.3 | NP_032813.3 | 2 | 387 | |||
GenBank mRNA | AB207233.1 | - | 2 | 261 | ||
AK145315.1 | - | 2 | 208 | |||
BB840023.1 | - | 2 | 207 | |||
BC034865.1 | - | 2 | 194 | |||
BC085244.1 | - | 2 | 211 | |||
BC100380.1 | - | 2 | 182 |