Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252437.1 | NP_001239366.1 | 5, 6 | 849 | ||
NM_029657.4 | NP_083933.1 | 5, 6 | 846 | |||
GenBank mRNA | AK011747.2 | - | 5, 6 | 639 | ||
AK088533.1 | - | 5, 6 | 629 | |||
AK129161.1 | - | 2, 3 | 257 | |||
AK150176.1 | - | 6, 7 | 682 | |||
AK153407.1 | - | 5, 6 | 670 | |||
BC046830.1 | - | 5, 6 | 695 | 6811606 |