Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003831.3 | NP_003822.2 | 13 | 2163 | ||
XM_011526242.1 | XP_011524544.1 | 12 | 2035 | |||
XM_011526243.1 | XP_011524545.1 | 13 | 2020 | |||
GenBank mRNA | AF013591.1 | - | 13 | 1639 | ||
AK057292.1 | - | 13 | 2163 | |||
AK303938.1 | - | 13 | 1651 | |||
AK304571.1 | - | 8 | 858 | |||
AK310783.1 | - | 6 | 788 | |||
BC028360.1 | - | 12 | 1791 | 4828351 | ||
BC039729.1 | - | 13 | 1669 | 5745005 | ||
BX640929.1 | - | 2 | 512 |